Avantor™

TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker™

Catalog number: 4103-02
Have Questions?
Avantor™

TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker™

Catalog number: 4103-02
Have Questions?
For laboratory, research, or manufacturing use.
 
Catalog Number
4103-02
Unit Size
Each
Quantity
1 kg
Packaging
White poly bottle, tamper-evident cap
Price (USD)
Full specifications
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity1 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingWhite poly bottle, tamper-evident cap
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
500 g
Packaging
White poly bottle, tamper-evident cap
Price (USD)
Full specifications
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity500 g
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingWhite poly bottle, tamper-evident cap
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
250 g
Packaging
White poly bottle, tamper-evident cap
Price (USD)
Full specifications
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity250 g
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingWhite poly bottle, tamper-evident cap
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
5 kg
Packaging
Polyethylene pail, screw top
Price (USD)
Full specifications
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity5 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPolyethylene pail, screw top
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
12 kg
Packaging
Polyethylene pail, screw top
Price (USD)
Full specifications
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity12 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPolyethylene pail, screw top
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
50 kg
Packaging
Polyethylene drum, open head
Price (USD)
Full specifications
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity50 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPolyethylene drum, open head
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
12 kg
Packaging
Pail, HDPE Flowmor™ package
Price (USD)
Full specifications
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity12 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPail, HDPE Flowmor™ package
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
Showing 7 of 7
Catalog NumberSpecificationsUnit SizeQuantityPackagingPrice (USD)
4103-02Full specifications
Each1 kgWhite poly bottle, tamper-evident capRequest A Quote
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity1 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingWhite poly bottle, tamper-evident cap
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
4103-01Full specifications
Each500 gWhite poly bottle, tamper-evident capRequest A Quote
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity500 g
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingWhite poly bottle, tamper-evident cap
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
4103-04Full specifications
Each250 gWhite poly bottle, tamper-evident capRequest A Quote
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity250 g
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingWhite poly bottle, tamper-evident cap
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
4103-06Full specifications
Each5 kgPolyethylene pail, screw topRequest A Quote
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity5 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPolyethylene pail, screw top
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
4103-07Full specifications
Each12 kgPolyethylene pail, screw topRequest A Quote
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity12 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPolyethylene pail, screw top
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
4103-08Full specifications
Each50 kgPolyethylene drum, open headRequest A Quote
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity50 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPolyethylene drum, open head
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
4103-F7Full specifications
Each12 kgPail, HDPE Flowmor™ packageRequest A Quote
Chemical Name or MaterialTRIS HCl
CAS1185-53-1
Melting Point150–152°C
ColorColorless
For Use With (Application)Pharma and Biopharma
Physical FormCrystalline
Quantity12 kg
Solution TypeBuffer
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPail, HDPE Flowmor™ package
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
GradeMolecular Biology Ultrapure
Unit SizeEach
Showing 7 of 7
For laboratory, research, or manufacturing use.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text