| 4103-04 | Full specifications | Each | 250 g | White poly bottle, tamper-evident cap | Request A Quote |
Chemical Name or Material | TRIS HCl | CAS | 1185-53-1 | Melting Point | 150–152°C | Color | Colorless | For Use With (Application) | Pharma and Biopharma | Physical Form | Crystalline | Quantity | 250 g | Solution Type | Buffer | Molecular Formula | NH2C(CH2OH)3·HCl | Packaging | White poly bottle, tamper-evident cap | Synonym | Tris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl | Molecular Weight (g/mol) | 157.6 | Percent Purity | ≥99.0% | Grade | Molecular Biology Ultrapure | Unit Size | Each |
|
| 4103-01 | Full specifications | Each | 500 g | White poly bottle, tamper-evident cap | Request A Quote |
Chemical Name or Material | TRIS HCl | CAS | 1185-53-1 | Melting Point | 150–152°C | Color | Colorless | For Use With (Application) | Pharma and Biopharma | Physical Form | Crystalline | Quantity | 500 g | Solution Type | Buffer | Molecular Formula | NH2C(CH2OH)3·HCl | Packaging | White poly bottle, tamper-evident cap | Synonym | Tris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl | Molecular Weight (g/mol) | 157.6 | Percent Purity | ≥99.0% | Grade | Molecular Biology Ultrapure | Unit Size | Each |
|
| 4103-02 | Full specifications | Each | 1 kg | White poly bottle, tamper-evident cap | Request A Quote |
Chemical Name or Material | TRIS HCl | CAS | 1185-53-1 | Melting Point | 150–152°C | Color | Colorless | For Use With (Application) | Pharma and Biopharma | Physical Form | Crystalline | Quantity | 1 kg | Solution Type | Buffer | Molecular Formula | NH2C(CH2OH)3·HCl | Packaging | White poly bottle, tamper-evident cap | Synonym | Tris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl | Molecular Weight (g/mol) | 157.6 | Percent Purity | ≥99.0% | Grade | Molecular Biology Ultrapure | Unit Size | Each |
|
| 4103-06 | Full specifications | Each | 5 kg | Polyethylene pail, screw top | Request A Quote |
Chemical Name or Material | TRIS HCl | CAS | 1185-53-1 | Melting Point | 150–152°C | Color | Colorless | For Use With (Application) | Pharma and Biopharma | Physical Form | Crystalline | Quantity | 5 kg | Solution Type | Buffer | Molecular Formula | NH2C(CH2OH)3·HCl | Packaging | Polyethylene pail, screw top | Synonym | Tris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl | Molecular Weight (g/mol) | 157.6 | Percent Purity | ≥99.0% | Grade | Molecular Biology Ultrapure | Unit Size | Each |
|
| 4103-07 | Full specifications | Each | 12 kg | Polyethylene pail, screw top | Request A Quote |
Chemical Name or Material | TRIS HCl | CAS | 1185-53-1 | Melting Point | 150–152°C | Color | Colorless | For Use With (Application) | Pharma and Biopharma | Physical Form | Crystalline | Quantity | 12 kg | Solution Type | Buffer | Molecular Formula | NH2C(CH2OH)3·HCl | Packaging | Polyethylene pail, screw top | Synonym | Tris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl | Molecular Weight (g/mol) | 157.6 | Percent Purity | ≥99.0% | Grade | Molecular Biology Ultrapure | Unit Size | Each |
|
| 4103-08 | Full specifications | Each | 50 kg | Polyethylene drum, open head | Request A Quote |
Chemical Name or Material | TRIS HCl | CAS | 1185-53-1 | Melting Point | 150–152°C | Color | Colorless | For Use With (Application) | Pharma and Biopharma | Physical Form | Crystalline | Quantity | 50 kg | Solution Type | Buffer | Molecular Formula | NH2C(CH2OH)3·HCl | Packaging | Polyethylene drum, open head | Synonym | Tris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl | Molecular Weight (g/mol) | 157.6 | Percent Purity | ≥99.0% | Grade | Molecular Biology Ultrapure | Unit Size | Each |
|
| 4103-F7 | Full specifications | Each | 12 kg | Pail, HDPE Flowmor™ package | Request A Quote |
Chemical Name or Material | TRIS HCl | CAS | 1185-53-1 | Melting Point | 150–152°C | Color | Colorless | For Use With (Application) | Pharma and Biopharma | Physical Form | Crystalline | Quantity | 12 kg | Solution Type | Buffer | Molecular Formula | NH2C(CH2OH)3·HCl | Packaging | Pail, HDPE Flowmor™ package | Synonym | Tris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl | Molecular Weight (g/mol) | 157.6 | Percent Purity | ≥99.0% | Grade | Molecular Biology Ultrapure | Unit Size | Each |
|