Paraffin, pure, wax, granular
Paraffin, pure, wax, granular
Paraffin, pure, wax, granular
Paraffin, pure, wax, granular
Thermo Scientific Chemicals

Paraffin, pure, wax, granular

Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure | CAS: 8002-74-2 | CnH2n+2 | 341.451 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
2 kg
Catalog number 416770100
Price (USD)
416.00
Each
In stock
Add to cart
Quantity:
10 kg
Request bulk or custom format
Price (USD)
416.00
Each
Add to cart
Chemical Identifiers
CAS8002-74-2
IUPAC Name1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one
Molecular FormulaC21H27NO3
InChI KeyJWHAUXFOSRPERK-UHFFFAOYNA-N
SMILESCCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Pellets
Infrared spectrumConforms
Congealing point58°C to 62°C
Ask our AI about this Product
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0448635Certificate of AnalysisJun 23, 2025416770100, 416770020
A0472270Certificate of AnalysisMay 17, 2025416770100, 416770020
A0467441Certificate of AnalysisNov 11, 2024416770100, 416770020
A0461881Certificate of AnalysisApr 11, 2024416770100, 416770020
A0455161Certificate of AnalysisAug 25, 2023416770100, 416770020
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection