Aluminum potassium sulfate dodecahydrate, 98+%, ACS reagent
Aluminum potassium sulfate dodecahydrate, 98+%, ACS reagent
Aluminum potassium sulfate dodecahydrate, 98+%, ACS reagent
Aluminum potassium sulfate dodecahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Aluminum potassium sulfate dodecahydrate, 98+%, ACS reagent

CAS: 7784-24-9 | AlKO8S2·12H2O | 474.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 423265000
Price (USD)
104.65
Special offer
Online exclusive
Ends: 26-Sep-2025
129.00
Save 24.35 (19%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
104.65
Special offer
Online exclusive
Ends: 26-Sep-2025
129.00
Save 24.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7784-24-9
IUPAC Namealuminium(3+) potassium dodecahydrate disulfate
Molecular FormulaAlH24KO20S2
InChI KeyGNHOJBNSNUXZQA-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration Complexometric98.0 to 102.0 %
Heavy metals (as Pb)=<10 ppm
Ammonium (NH4)=<50 ppm
Appearance (Color)White
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470589Certificate of AnalysisMay 17, 2025423260050, 423265000, 423260025
A0458136Certificate of AnalysisNov 27, 2023423260050, 423265000, 423260025
A0453738Certificate of AnalysisAug 25, 2023423260050, 423265000, 423260025
A0451513Certificate of AnalysisMay 02, 2023423260050, 423265000, 423260025
A0376628Certificate of AnalysisDec 13, 2022423260050, 423265000, 423260025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection