Persulfato potásico, +99 %, reactivo ACS, Thermo Scientific Chemicals
Persulfato potásico, +99 %, reactivo ACS, Thermo Scientific Chemicals
Persulfato potásico, +99 %, reactivo ACS, Thermo Scientific Chemicals
Persulfato potásico, +99 %, reactivo ACS, Thermo Scientific Chemicals
Persulfato potásico, +99 %, reactivo ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Persulfato potásico, +99 %, reactivo ACS, Thermo Scientific Chemicals

CAS: 7727-21-1 | K2O8S2 | 270.3 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
2.5 kg
500 g
Envase:
Frasco de plástico
Número de catálogo 424185000
Precio (EUR)
44,46
Precio exclusivo en nuestra web
53,60
Ahorro 9,14 (17%)
Each
-
Añadir al carro de la compra
Cantidad:
500 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Precio (EUR)
44,46
Precio exclusivo en nuestra web
53,60
Ahorro 9,14 (17%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS7727-21-1
IUPAC Namedipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaK2O8S2
InChI KeyUSHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Titration with KMnO4>=99.0 %
Heavy metals (as Pb)=<0.001 %
Insoluble matter=<0.005 %
Chlorine compounds (as Cl)=<0.001 %
Iron (Fe)=<5 ppm
Ver más
Ask our AI about this Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Reactivo de oxidación
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0472778Certificate of Analysis23 may 2025424185000, 424180025
A0467241Certificate of Analysis07 nov 2024424185000, 424180025
A0463099Certificate of Analysis11 jun 2024424185000, 424180025
A0458564Certificate of Analysis12 dic 2023424185000, 424180025
A0435142Certificate of Analysis04 abr 2023424185000, 424180025
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Oxidizing solids
Category 3
Acute oral toxicity
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Respiratory Sensitization
Category 1
Skin Sensitization
Category 1
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H302 - Harmful if swallowed

H315 - Causes skin irritation

H317 - May cause an allergic skin reaction

H319 - Causes serious eye irritation

H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

H335 - May cause respiratory irritation

Physical Hazards

H272 - May intensify fire; oxidizer

Precautionary Statements

P220 - Keep away from clothing and other combustible materials

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P342 + P311 - If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection