Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.
Diameter (Metric)Top | 62 mm |
Capacity (Metric) | 62 mL |
Diameter (Metric) Stem | 14 mm |
Material | Polypropylene |
Height (Metric) | 66 mm |
Autoclavable | Yes |
Description | Funnel Utility PP 70mm |
Length (Metric) Stem | 21mm |
Quantity | 12/Pk.; 288/Cs. |
Unit Size | Pack of 12 |
Diameter (Metric)Top | 62 mm |
Capacity (Metric) | 62 mL |
Diameter (Metric) Stem | 14 mm |
Material | Polypropylene |
Height (Metric) | 66 mm |
Autoclavable | Yes |
Description | Funnel Utility PP 70mm |
Length (Metric) Stem | 21mm |
Quantity | 12/Pk.; 288/Cs. |
Unit Size | Case of 288 |
Diameter (Metric)Top | 98 mm |
Capacity (Metric) | 270 mL |
Diameter (Metric) Stem | 18 mm |
Material | Polypropylene |
Height (Metric) | 117 mm |
Autoclavable | Yes |
Description | Funnel Utility PP 108mm |
Length (Metric) Stem | 37 mm |
Quantity | 12/Pk.; 144/Cs. |
Unit Size | Case of 144 |
Diameter (Metric)Top | 98 mm |
Capacity (Metric) | 270 mL |
Diameter (Metric) Stem | 18 mm |
Material | Polypropylene |
Height (Metric) | 117 mm |
Autoclavable | Yes |
Description | Funnel Utility PP 108mm |
Length (Metric) Stem | 37 mm |
Quantity | 12/Pk.; 144/Cs. |
Unit Size | Pack of 12 |
Diameter (Metric)Top | 150 mm |
Capacity (Metric) | 1059 mL |
Diameter (Metric) Stem | 48 mm |
Material | HDPE |
Height (Metric) | 175 mm |
Autoclavable | No |
Description | Funnel Utility HDPE 162mm |
Length (Metric) Stem | 66 mm |
Quantity | 12/Pk.; 72/Cs. |
Unit Size | Case of 72 |
Diameter (Metric)Top | 150 mm |
Capacity (Metric) | 1059 mL |
Diameter (Metric) Stem | 48 mm |
Material | HDPE |
Height (Metric) | 175 mm |
Autoclavable | No |
Description | Funnel Utility HDPE 162mm |
Length (Metric) Stem | 66 mm |
Quantity | 12/Pk.; 72/Cs. |
Unit Size | Pack of 12 |
Catalog Number | Specifications | Unit Size | Diameter (Metric)Top | Capacity (Metric) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4256-0234PK also known as 4256-0234 | Pack of 12 | 62 mm | 62 mL | 71.10 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4256-0234 | Case of 288 | 62 mm | 62 mL | 1,447.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4256-0414 | Case of 144 | 98 mm | 270 mL | 887.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4256-0414PK also known as 4256-0414 | Pack of 12 | 98 mm | 270 mL | 87.30 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4256-0638 | Case of 72 | 150 mm | 1059 mL | 1,556.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4256-0638PK also known as 4256-0638 | Pack of 12 | 150 mm | 1059 mL | 305.80 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|