Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers
Thermo Scientific Chemicals

Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers

CAS: 34590-94-8 | C7H16O3 | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 428200010
Price (USD)
53.65
Online Exclusive
58.90
Save 5.25 (9%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
58.90
Save 5.25 (9%)
Each
Add to cart
Chemical Identifiers
CAS34590-94-8
IUPAC Name2-(2-methoxypropoxy)propan-1-ol
Molecular FormulaC7H16O3
InChI KeyCUDYYMUUJHLCGZ-UHFFFAOYNA-N
SMILESCOC(C)COC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 % sum of isomers
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470606Certificate of AnalysisMay 17, 2025428200050, 428200010
A0466999Certificate of AnalysisOct 22, 2024428200050, 428200010
A0463194Certificate of AnalysisJun 17, 2024428200050, 428200010
A0455624Certificate of AnalysisAug 25, 2023428200050, 428200010
A0451305Certificate of AnalysisMay 02, 2023428200050, 428200010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection