Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Upgrade to Platinum II Taq Hot-Start DNA Polymerase for universal annealing and a 4X faster cycling protocol.
Applied Biosystems™

AmpliTaq Gold™ DNA Polymerase with Gold Buffer and MgCl2

Catalog number: 4311816
Have Questions?
Applied Biosystems™

AmpliTaq Gold™ DNA Polymerase with Gold Buffer and MgCl2

Catalog number: 4311816
Have Questions?
Catalog Number
4311816
Unit Size
1,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
3,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43118161,000 units
-
4311806250 units
-
43118203,000 units
-
43118185,000 units
-
431774225,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

AmpliTaq Gold™ DNA Polymerase is a chemically modified form of AmpliTaq™ DNA Polymerase requiring thermal activation.

Features of this enzyme:

  • Automated chemical hot-start enzyme for increased specificity, sensitivity, and yield
  • Time-released thermal activation improves sensitivity in low copy number amplifications
  • Successful multiplex PCR saves time and reagents
  • Includes GeneAmp™ 10X PCR Gold Buffer with MgCl2

Hot-start activation

The modified enzyme is provided in an inactive state. Upon thermal activation, the modifier is permanently released, regenerating active enzyme. The resulting hot-start PCR amplification provides increased sensitivity, specificity, and yield over conventional PCR techniques.

Higher specificity, higher yield

AmpliTaq Gold™ DNA Polymerase can be activated partially or completely in a pre-PCR heat step, or can be allowed to activate slowly in a time-released manner during the denaturation steps of thermal cycling. With or without a limited up-front heat activation step, active enzyme is released slowly during thermal cycling to match template concentration and increase specificity. The yield of specific product increases because reactants are not wasted in the formation of unintended products. Because AmpliTaq Gold™ DNA Polymerase is a chemical hot-start enzyme, there is no worry of biological contamination.

GeneAmp™ 10X PCR Gold Buffer is formulated to provide flexible, efficient activation of AmpliTaq Gold™ DNA Polymerase, resulting in a highly specific, robust PCR amplification. The ionic strength and the pH of GeneAmp™ 10X PCR Gold Buffer have been optimized to provide a wider activation temperature and time range when used in conjunction with AmpliTaq Gold™ DNA Polymerase.

AmpliTaq Gold™ 360 DNA Polymerase for superior performance

Also available is AmpliTaq Gold™ 360 DNA Polymerase for even better PCR performance. Compared to AmpliTaq Gold™ DNA Polymerase, AmpliTaq Gold™ 360 DNA Polymerase is purified by an additional proprietary separation process to eliminate contaminating bacterial DNA sequences from the enzyme preparation. When used with the enhanced 360 Gold Buffer, this ultra-pure enzyme, in addition to its hot-start capabilities, reduces false positive results, amplifies low-level target sequences, and promotes the amplification of a variety of templates. Also included with this product is a 360 GC Enhancer to work through challenging GC-rich templates.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Tube
GC-Rich PCR Performance
Low
Polymerase
AmpliTaq Gold DNA Polymerase
Reaction Speed
Standard
Exonuclease Activity
5' - 3'
Product Type
DNA Polymerase
Quantity
1000 Units
Shipping Condition
Dry Ice
For Use With (Application)
Hot-start PCR
Concentration
5 U/μL
Fidelity (vs. Taq)
1X
Hot Start
Built-In Hot Start
No. of Reactions
800
Overhang
3'-A
Reaction Format
Separate Components
Size (Final Product)
5 kb or less
Starting Material
DNA

Contents & Storage

• AmpliTaq Gold DNA Polymerase (5 U/μL), 1 x 200 μL
• GeneAmp 10X Gold Buffer, 4 x 1.5 mL
• 25 mM MgCl2, 4 x 1.5 mL

Store at -15°C to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text