Applied Biosystems™

TaqMan™ MGB Probe

Catalog number: 4316032
Have Questions?
Applied Biosystems™

TaqMan™ MGB Probe

Catalog number: 4316032
Have Questions?
Catalog Number
4316032
Unit Size
50,000 pmol
Price (USD)
Catalog Number
Unit Size
6,000 pmol
Price (USD)
Catalog Number
Unit Size
20,000 pmol
Price (USD)
Catalog NumberUnit SizePrice (USD)
431603250,000 pmol
43160346,000 pmol
431603320,000 pmol
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Applied Biosystems TaqMan MGB (minor groove binder) probes are dual-labeled probes used for real-time PCR applications using TaqMan chemistry. TaqMan MGB Probes incorporate a 5' fluorescent reporter dye and a 3' nonfluorescent quencher (NFQ). The NFQ offers the advantage of lower background signal, which results in better precision in quantitation. Custom TaqMan MGB probes are HPLC-purified and available with FAM, VIC, TET, and NED reporter dyes.

Shorter, more specific probes
TaqMan MGB probes include a MGB moiety at the 3’ end that increases the melting temperature (Tm) of the probe and stabilizes probe/target hybrids. This means that TaqMan MGB probes can be significantly shorter than traditional probes, providing better sequence discrimination and flexibility to accommodate more targets.

Nonfluorescent quencher (NFQ) maximizes sensitivity
TaqMan MGB probes incorporate an NFQ to absorb (quench) signal from the fluorescent dye label at the other end of the probe. The properties of the NFQ combined with the short length of the MGB probe result in lower background signal than with non-MGB NFQ probes. Lower background means increased sensitivity and precision in your data.

Guaranteed yield
The unit size you select is based on the minimum amount we will deliver, not on synthesis starting amount.

Approximate ship time
4–6 days in North America, 4-7 days in Europe

We also offer:
• Applied Biosystems custom primers and other types of TaqMan probes. Learn more ›
• TaqMan MGB probes and primers available as predesigned and pre-formulated TaqMan Assays. Learn more ›
• Commercial manufacturing products and services, including OEM and GMP oligo manufacturing. Learn more ›

Need a different dye or concentration or a larger lot size? The TaqMan Custom Assay and Oligo Service offers custom products that you can tailor to meet your experimental needs. Learn more ›

Specifications

3'Primer Modification
MGB-NFQ
5'Primer Modification
FAM, NED, TET, VIC
Form
Liquid
Format
Tube
For Use With (Equipment)
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
Green Features
Sustainable packaging
Internal Probe Modification
None
Label or Dye
NED, TET
Product Line
TaqMan™
Product Type
TaqMan Probe
Purification Method
HPLC
Purity or Quality Grade
HPLC
Quantity
50,000 pmol Scale
Shipping Condition
Room Temperature
For Use With (Application)
Real Time PCR (qPCR)

Contents & Storage

1 tube containing a dual-labeled probe in solution.

Store at -15°C to -25°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text