Applied Biosystems™

TaqMan™ Gene Expression Master Mix

Catalog number: 4369510
Have Questions?
Applied Biosystems™

TaqMan™ Gene Expression Master Mix

Catalog number: 4369510
Have Questions?
Catalog Number
4369510
Unit Size
5 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43695105 x 5 mL
-
436954210 x 5 mL
-
43700481 x 1 mL
-
43695142 x 5 mL
-
43690161 x 5 mL
-
43700741 x 50 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Get precise and reliable real-time qPCR quantification over nine logs of linear dynamic range with TaqMan Gene Expression Master Mix. TaqMan Gene Expression Master Mix is an optimized 2X mix that contains all of the components, excluding the template and primers, for sensitive detection down to one copy of target. Extended benchtop stability provides consistent results for automated liquid-handling systems, making it the choice for high-throughput sample processing, overnight or over the weekend. TaqMan Gene Expression Master Mix has been validated with TaqMan gene expression assays and Applied Biosystems real-time systems, providing precise quantification for a variety of gene expression–based real-time qPCR applications, including:
• Rare transcript detection
• Gene expression analysis
• Pathogen detection and viral load quantification
• Co-amplification of two targets
• Specific detection of gene family members

TaqMan Gene Expression Master Mix contains AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), for hot start activation and improved detection of bacterial targets, a blend of dNTPs with dTTP/dUTP and uracil-DNA glycosylase (UDG) to minimize carry-over PCR contamination, and a passive internal reference based on proprietary ROX dye for superb precision on Applied Biosystems real-time PCR instruments.

Alternative product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Gene Expression Master Mix and added additional capabilities for your gene expression analysis.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
PCR Method
qPCR
Polymerase
AmpliTaq Gold DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7500 System, 7900HT System, Applied Biosystems StepOnePlus™ Fast Real-Time PCR SystemStepOne™, Standard Mode, StepOnePlus™, Standard Mode
Passive Reference Dye
ROX (Pre-mixed)
Product Line
TaqMan™
Purity or Quality Grade
UP (Ultra Pure)
Quantity
5 x 5 mL
For Use With (Application)
Gene Expression
No. of Reactions
1000 Reactions
Sample Type
DNA (Genomic), cDNA
Volume
5 x 5 mL

Contents & Storage

5 bottles (5 mL each) 2X TaqMan™ Gene Expression Master Mix sufficient for 1000 reactions. Store at 2°C to 8°C.


TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase (Ultra Pure), Uracil-DNA glycosylase, dNTPs (with dUTP), ROX™ Passive Reference, and optimized buffer components.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text