Calcium carbonate, 99.999%, (trace metal basis)
Calcium carbonate, 99.999%, (trace metal basis)
Calcium carbonate, 99.999%, (trace metal basis)
Calcium carbonate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Calcium carbonate, 99.999%, (trace metal basis)

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 437190250
Price (USD)
154.65
Special offer
Online exclusive
182.00
Save 27.35 (15%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
154.65
Special offer
Online exclusive
182.00
Save 27.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Complexometric39 to 42 %
Total trace metal impurities=<10 ppm
Appearance (Form)Powder
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470743Certificate of AnalysisMay 17, 2025437190050, 437190250
A0468829Certificate of AnalysisJan 31, 2025437190050, 437190250
A0461875Certificate of AnalysisApr 11, 2024437190050, 437190250
A0446867Certificate of AnalysisJan 19, 2023437190050, 437190250
A0407448Certificate of AnalysisDec 13, 2022437190050, 437190250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection