Thermo Scientific Chemicals

Sodium hexanoate, 99%, Thermo Scientific Chemicals

Catalog number: 437540250
25 g, Each
Thermo Scientific Chemicals

Sodium hexanoate, 99%, Thermo Scientific Chemicals

Catalog number: 437540250
25 g, Each
Quantity
Catalog number: 437540250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10051-44-2
IUPAC Name
sodium hexanoate
Molecular Formula
C6H11NaO2
InChI Key
UDWXLZLRRVQONG-UHFFFAOYSA-M
SMILES
[Na+].CCCCCC([O-])=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Loss on drying
=<2 %
Titration with HClO4
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text