alpha-Ketoglutaric acid sodium salt, 98%
alpha-Ketoglutaric acid sodium salt, 98%
alpha-Ketoglutaric acid sodium salt, 98%
alpha-Ketoglutaric acid sodium salt, 98%
Thermo Scientific Chemicals

alpha-Ketoglutaric acid sodium salt, 98%

CAS: 22202-68-2 | C5H5NaO5 | 168.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 439350050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS22202-68-2
IUPAC Namedisodium 2-oxopentanedioate
Molecular FormulaC5H4Na2O5
InChI KeyYBGBJYVHJTVUSL-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)CCC(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<1.0 %
UVat 430 nm A: =<0.200
Appearance (Color)White
Titration with HClO4>=97.5 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
I am getting lower yield than I expected with the ExpiCHO expression system. What should I do to improve my results?
I don't have ExpiCHO-S cells. Can Expifectamine CHO transfection reagent and enhancer be used with other CHO cell lines than the ExpiCHO-S cell line?
What type of Rabbit IgG does the Antibody-Expressing Positive Control Vector (Cat. No. A14662) express?
Do I need to change the media after the ExpiFectamine CHO transfection?
How does the protein expression yield of the ExpiCHO expression system compare to FreeStyle CHO or Expi293 expression systems?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
345418Certificate of AnalysisOct 16, 2024439351000, 439350250, 439350050
345073Certificate of AnalysisMay 02, 2023439351000, 439350250, 439350050
341017Certificate of AnalysisDec 12, 2022439351000, 439350250, 439350050
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection