Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Switch to Thermo Scientific DreamTaq DNA Polymerases for better results and added convenience.
Applied Biosystems™

AmpliTaq™ 360 DNA Polymerase

Catalog number: 4398828
Have Questions?
Applied Biosystems™

AmpliTaq™ 360 DNA Polymerase

Catalog number: 4398828
Have Questions?
Catalog Number
4398828
Unit Size
1,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 x 1,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43988281,000 units
-
4398818250 units
-
43988955,000 units
-
439889725 x 1,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. Compared to the original AmpliTaq DNA polymerase, AmpliTaq 360 polymerase is purified by an additional proprietary separation process which reduces contaminating bacterial DNA sequences from the enzyme preparation. This ultra-pure enzyme reduces false positive results, amplifies low-level target sequences, and when combined with the proprietary AmpliTaq 360 Buffer Kit, promotes the amplification of a variety of templates including those from bacterial and human genomes.

• Optimized for the broadest range of targets—from everyday to challenging
• Unmatched sensitivity and yield
• Market-leading GC enhancer for robust amplification of GC-rich sequences
• Achieves the highest quality sequencing data

For superior PCR performance, we recommend DreamTaq DNA Polymerase.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Stand-alone enzyme
GC-Rich PCR Performance
Medium
Polymerase
AmpliTaq 360 DNA Polymerase
Reaction Speed
Standard
Exonuclease Activity
5' - 3'
Green Features
Sustainable packaging
Product Line
AmpliTaq™
Product Type
DNA Polymerase
Quantity
1000 Units
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice
Concentration
5 U/μL
Fidelity (vs. Taq)
1 X
Hot Start
No
No. of Reactions
800
Overhang
3'-A
Reaction Format
Separate Components
Size (Final Product)
5 kb or less
Starting Material
DNA

Contents & Storage

AmpliTaq™ 360 DNA Polymerase is supplied at 5U⁄μL with 10X AmpliTaq™ 360 PCR Buffer, 25mM M2Cl2 , and 360 GC Enhancer. Typically, 1.5mL of each of AmpliTaq 360 Buffer, 25mM MgCl2 , and 360 GC Enhancer is supplied with each 250U of enzyme.
• Storage Buffer Enzyme is supplied in 20 mM Tris, pH 9.0, 100 mM KCl, 0.1 mM EDTA, 1 mM DTT, 0.5% (w⁄v) Tween 20,
50% (v⁄v) glycerol
• Source: Recombinant, modified form of the Thermus aquaticus DNA polymerase gene expressed in E. coli

All solutions are prepared in deionized, filter-sterilized water. When stored at -15 to -25 °C, this product maintains performance at least until the expiration date on the tube label. Before use, thaw frozen components to room temperature and mix well.

Product is guaranteed up to 12 months as indicated by the expiration date.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text