Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
4427807 | 1 kit |
This MicroSEQ™ ID Purification Combo Kit includes reagents for post-PCR and post-sequencing steps in the MicroSEQ™ ID microbial identification workflow. It contains 2 tubes of ExoSAP-IT™ Reagent for PCR clean-up and 36 MicroSEQ™ ID Sequencing Clean-up Cartridges.
ExoSAP-IT™ Reagent: this reagent is used for quick post-PCR cleanup for downstream applications such as sequencing. It allows for removal of unconsumed dNTPs and primers remaining in the PCR mix following amplification.
ExoSAP-IT™ Reagent can be used downstream of the following PCR amplification kits:
MicroSEQ™ ID Sequencing Clean-up Cartridges: these cartridges are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrix used in the cartridges provide a convenient and simple way to remove dye terminators, dNTPs, and salts, as well as residual primers from cycle sequencing reactions.
The MicroSEQ™ ID Sequencing Cleanup Cartridges can be used downstream of the following sequencing kits:
Notice to Purchaser: The purchase of this product conveys to the purchaser the limited, non-transferable right to use the purchased amount of the product (a) to perform internal research for the sole benefit of the purchaser; and (b) for environmental testing, quality control/quality assurance testing, food and agricultural testing, including reporting results of purchaser's activities in environmental testing, quality control/quality assurance testing, food and agricultural testing for a fee or other commercial consideration. No other right is hereby granted expressly, by implication, or by estoppel. This product is for environmental testing, quality control/ quality assurance testing, food and agricultural testing and research purposes only. The purchase of this product does not grant the purchaser any additional rights, including (without limitation) the right to transfer or resell the product in any form or the right to use the product as a therapeutic agent or diagnostics test component. For information on obtaining additional rights, please contact outlicensing@thermofisher.com, Licensing and Commercial Supply, Thermo Fisher Scientific, 5823 Newton Drive, Carlsbad, CA, 92008, United States.
'Share catalog number, name or link