n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.
Thermo Scientific™

n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.

CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445720025
Price (USD)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water soluble titr. acid=<0.0003 meq/g
UVat 220 nm A: =<0.07
UVat 240 nm A: =<0.01
UVat 260 nm A: =<0.005
Density(20/20°C) 0.659 to 0.663
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2539324Certificate of AnalysisJun 09, 2025445720010, 445720025
2534196Certificate of AnalysisJun 09, 2025445720010, 445720025
2532504Certificate of AnalysisMay 24, 2025445720010, 445720025
2524702Certificate of AnalysisJan 29, 2025445720010, 445720025
2496512Certificate of AnalysisDec 23, 2024445720010, 445720025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection