1x1 image pixel for data collection
Pierce™ Antibody Clean-up Kit
Pierce™ Antibody Clean-up Kit
Thermo Scientific™

Pierce™ Antibody Clean-up Kit

The Thermo Scientific Pierce Antibody Clean-Up Kit is for removing BSA, gelatin and other stabilizer proteins from antibody stock solutionsRead more
Have Questions?
Catalog NumberQuantity
4460010 Reactions
Catalog number 44600
Price (USD)
349.65
Online Exclusive
395.00
Save 45.35 (11%)
Each
In stock
Add to cart
Quantity:
10 Reactions
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
349.65
Online Exclusive
395.00
Save 45.35 (11%)
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific Pierce Antibody Clean-Up Kit is for removing BSA, gelatin and other stabilizer proteins from antibody stock solutions before amine-based chemical labeling and conjugation procedures.

Features of the Antibody Clean-Up Kit:

Fast—removes carrier protein from antibody solutions in half the time required for purifying antibodies through Protein A or G purification procedures
Gentle no need to neutralize or desalt to mitigate the effects of harsh elution buffers
Complete—supplied desalting columns provide for easy buffer exchange into the required purification buffer or quick exchange into final buffer of choice
Economical—sufficient reagents to perform 10 antibody clean-up reactions

This is a complete and easy-to-use kit for removing BSA and gelatin stabilizing proteins (up to 1%) from IgG samples. The protocol uses Thermo Scientific Melon Gel to bind these frequently used storage proteins, allowing the antibody to flow through in a purified form. The antibody is recovered in an amine-free and carboxyl-free buffer, ready for use in downstream applications such as biotinylation, fluorescent labeling, crosslinking, immobilization and enzymatic fragmentation (e.g., Fab preparation).

Bovine serum albumin (BSA) and gelatin are often added to purified antibody solutions as stabilizers for long-term storage. Inclusion of 0.2 to 1% (2 to 10 mg/mL) BSA or gelatin helps stabilize dilute antibody solutions. Generally these added proteins do not interfere with immunodetection methods; however, their presence in solution does interfere with amine-reactive conjugation chemistries that are the basis for most biotinylation, fluorescent dye labeling, antibody fragmentation and covalent immobilization methods. For such methods, the antibody must be isolated from the stabilizing proteins, and the Antibody Clean-up Kit accomplishes this efficiently.

Related Products
Melon™ Gel IgG Spin Purification Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Purification TargetIgG Antibodies
Quantity10 Reactions
Column TypeProprietary Resin, Affinity
FormatKit, Microcentrifuge Spin Column
Product LinePierce™
Product TypeAntibody Clean-up Kit
Stationary PhaseProprietary Ligand
Unit SizeEach
Contents & Storage
Sufficient For: Purifying 10 x 0.5 mL IgG samples• Melon Gel IgG Purification Support, 1.2 mL
• Melon Gel Purification Buffer, 100 mL
• Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4°C
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Will glycerol pose a problem when purifying antibodies with the Pierce Antibody Clean-up Kit (Cat. No. 44600)?

When using the Pierce Antibody Clean-up Kit, glycerol may pose a problem for the spin desalting columns for buffer exchange, but not for the Melon Gel itself. If glycerol concentrations are high, the solution becomes viscous enough to exert a frictional drag during spin column chromatography and the performance and yield may suffer. Please find a technical tip on using Melon Gel for cleaning up BSA or gelatin from commercial antibodies.

If the glycerol concentration is higher than 10%, dialysis may be considered to exchange the sample into the Melon Gel buffer. We have some dialysis devices suitable for small volume dialysis, e.g., the Slide-A-Lyzer MINI devices: see here. Alternatively, some of the smaller Slide-A-Lyzer dialysis cassettes can be used: see here.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

I want to use the Antibody Clean-Up Kit (Cat. No. 44600) with a purified antibody that has a "protein based stabilizer". Do I need to first perform the Zeba desalting step A in the kit, or can I proceed directly to the BSA/Gelatin Removal step B?

The Zeba desalting step A is necessary as it results in a buffer exchange into the Melon Gel Purification Buffer. Having the antibody in this buffer is critical for the functionality of the Melon Gel used to remove protein.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

I typically use BSA or gelatin to store my purified antibody solution for long-term storage. I'm worried that it will interfere with my immunodetection experiment. Is there a way to remove BSA or gelatin from my antibody solution?

The Pierce Antibody Clean-up Kit (Cat. No. 44600) was specifically designed for removal of amine containing molecules from IgG. Please view this Tech Tip (https://tools.thermofisher.com/content/sfs/brochures/TR0055-Melon-Gel-for-BSA.pdf) for additional strategies on how to remove BSA and gelatin from antibody solutions.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3211964Certificate of AnalysisMay 12, 202544600
ZK402190Certificate of AnalysisDec 16, 202444600
ZK402190ACertificate of AnalysisDec 16, 202444600
ZH395953ACertificate of AnalysisSep 04, 202444600
ZH395953Certificate of AnalysisSep 04, 202444600
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link