Calcium phosphate, 96%, pure, mixture of Calcium phosphates
Calcium phosphate, 96%, pure, mixture of Calcium phosphates
Calcium phosphate, 96%, pure, mixture of Calcium phosphates
Calcium phosphate, 96%, pure, mixture of Calcium phosphates
Thermo Scientific Chemicals

Calcium phosphate, 96%, pure, mixture of Calcium phosphates

CAS: 7758-87-4 | Ca3O8P2 | 310.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 446390010
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7758-87-4
IUPAC Nametricalcium diphosphate
Molecular FormulaCa3O8P2
InChI KeyQORWJWZARLRLPR-UHFFFAOYSA-H
SMILES[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Complexometric>=95 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469286Certificate of AnalysisFeb 04, 2025446390050, 446390010
A0462616Certificate of AnalysisMay 07, 2024446390050, 446390010
A0457672Certificate of AnalysisNov 09, 2023446390050, 446390010
A0454550Certificate of AnalysisAug 25, 2023446390050, 446390010
A0392821Certificate of AnalysisDec 14, 2022446390050, 446390010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection