Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Carbon disulfide, 99.9%, ACS reagent
Thermo Scientific Chemicals

Carbon disulfide, 99.9%, ACS reagent

CAS: 75-15- | CS2 | 76.13 g/mol
Have Questions?
Catalog NumberChemical Name or Material
447035000Carbon disulfide
Catalog number 447035000
Price (USD)
62.65
Special offer
Online exclusive
73.70
Save 11.05 (15%)
Each
-
Add to cart
Chemical Name or Material:
Carbon disulfide
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
73.70
Save 11.05 (15%)
Each
Add to cart
Chemical Identifiers
CAS75-15-0
IUPAC Namemethanedithione
Molecular FormulaCS2
InChI KeyQGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILESS=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.05 %
Appearance (Form)Liquid
Sulfur dioxide (SO2)=<2.5 ppm
Residue after evaporation=<0.002 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0466897Certificate of AnalysisNov 11, 2024447035000
A0462124Certificate of AnalysisApr 19, 2024447035000
A0451439Certificate of AnalysisMay 02, 2023447035000
A0392371Certificate of AnalysisDec 14, 2022447035000
A0398930Certificate of AnalysisDec 12, 2022447035000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection