1x1 image pixel for data collection
HRM Software Module for QuantStudio™ 6 and 7 Systems
Product Image
Applied Biosystems™
HRM Software Module for QuantStudio™ 6 and 7 Systems
This software module expands the functionality of the QuantStudio™ 6 Flex and QuantStudio™ 7 Flex real-time PCR system software, allowingRead more
Have Questions?
Catalog NumberQuantity
448639510 licenses
Catalog number 4486395
Price (USD)
5,850.00
Each
In stock
Add to cart
Quantity:
10 licenses
Price (USD)
5,850.00
Each
Add to cart
Ask our AI about this Product
This software module expands the functionality of the QuantStudio™ 6 Flex and QuantStudio™ 7 Flex real-time PCR system software, allowing high resolution melting (HRM) analysis to be performed on PCR amplicons.
High Resolution Melt (HRM) analysis is a new, post-PCR analysis method used for identifying genetic variation in nucleic acid sequences. Simple and fast, this method is based on PCR melt (dissociation) curve techniques and is enabled by the recent availability of improved double-stranded DNA (dsDNA)-binding dyes along with next-generation real-time PCR instrumentation and analysis software. HRM analysis can discriminate DNA sequences based on their composition, length, GC content, or strand complementarity.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)QuantStudio™ 6 Flex, QuantStudio™ 7
Product LineQuantStudio™
Quantity10 licenses
Software TypeHRM Analysis Software
Product TypeSoftware Module
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What is the last software version for the ViiA7 Software?
ViiA7 Software v1.2.4 was the last version of the ViiA7 Software released. Currently, the ViiA7 Real-Time PCR System can be controlled using the QuantStudio Software. To obtain the most recent copy of the QuantStudio Software for performing experiments on the ViiA7 Real-Time PCR System, please visit the instrument software download page, https://www.thermofisher.com/us/en/home/technical-resources/software-downloads/applied-biosystems-viia-7-real-time-pcr-system.html.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection