Thermo Scientific™
Finnpipette™ F2 Fixed Volume Single-Channel Pipettes
Catalog number: 4652050
Have Questions?
Thermo Scientific™
Finnpipette™ F2 Fixed Volume Single-Channel Pipettes
Catalog number: 4652050
Have Questions?
Use Thermo Scientific™ Finnpipette™ F2 Fixed Volume Single-Channel Pipettes for highly demanding applications where robustness and durability are essential. With a sleek, rugged design, these pipettes are built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Fully autoclavable.
Catalog Number
4652050
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
100 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)100 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
1 μL
Compatible Tips
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)1 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
5 μL
Compatible Tips
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)5 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
10 μL
Compatible Tips
Finntip 250, Filter 10, Flex 200, Flex Filter 30
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 10, Flex 200, Flex Filter 30
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)10 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
25 μL
Compatible Tips
Finntip 250, Filter 30, Flex 200, Flex Filter 30
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 30, Flex 200, Flex Filter 30
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)25 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
50 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)50 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
250 μL
Compatible Tips
Finntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)250 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
500 μL
Compatible Tips
Finntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
ModelFinnPipette™ F2
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)500 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
1000 μL
Compatible Tips
Finntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)1000 μL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
FinnpipetteTM F2 fixed volume
Volume (Metric)
2000 μL
Compatible Tips
Finntip 5mL, Filter 5mL
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 5mL, Filter 5mL
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
ModelFinnPipette™ F2
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)2000 μL
Unit SizeEach
Showing 10 of 15Show more
Catalog NumberSpecificationsUnit SizeDescriptionVolume (Metric)Compatible TipsPrice (USD)AvailabilityQuantity
4652050Full specifications
EachFinnpipetteTM F2 fixed volume100 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)100 μL
Unit SizeEach
4652000Full specifications
EachFinnpipetteTM F2 fixed volume1 μLFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
-
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)1 μL
Unit SizeEach
4652010Full specifications
EachFinnpipetteTM F2 fixed volume5 μLFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
-
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)5 μL
Unit SizeEach
4652020Full specifications
EachFinnpipetteTM F2 fixed volume10 μLFinntip 250, Filter 10, Flex 200, Flex Filter 30
-
Compatible TipsFinntip 250, Filter 10, Flex 200, Flex Filter 30
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)10 μL
Unit SizeEach
4652030Full specifications
EachFinnpipetteTM F2 fixed volume25 μLFinntip 250, Filter 30, Flex 200, Flex Filter 30
-
Compatible TipsFinntip 250, Filter 30, Flex 200, Flex Filter 30
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)25 μL
Unit SizeEach
4652040Full specifications
EachFinnpipetteTM F2 fixed volume50 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)50 μL
Unit SizeEach
4652060Full specifications
EachFinnpipetteTM F2 fixed volume250 μLFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
-
Compatible TipsFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)250 μL
Unit SizeEach
4652070Full specifications
EachFinnpipetteTM F2 fixed volume500 μLFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
-
Compatible TipsFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
ModelFinnPipette™ F2
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)500 μL
Unit SizeEach
4652080Full specifications
EachFinnpipetteTM F2 fixed volume1000 μLFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
-
Compatible TipsFinntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)1000 μL
Unit SizeEach
4652090Full specifications
EachFinnpipetteTM F2 fixed volume2000 μLFinntip 5mL, Filter 5mL
-
Compatible TipsFinntip 5mL, Filter 5mL
DescriptionFinnpipetteTM F2 fixed volume
AutoclavableAutoclavable
MaterialPVDF
ModelFinnPipette™ F2
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)2000 μL
Unit SizeEach
Showing 10 of 15Show more
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.