Thermo Scientific™
Finnpipette™ F2 Multichannel Pipettes
Catalog number: 4662060
Have Questions?
Thermo Scientific™
Finnpipette™ F2 Multichannel Pipettes
Catalog number: 4662060
Have Questions?
Use the Thermo Scientific™ Finnpipette™ F2 Multichannel Pipette for highly demanding applications where robustness and durability are essential. With a sleek, rugged design, this pipette is built with PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Fully autoclavable and available in 8-, 12- and 16-channel models with a selection of volume ranges. In the low volume models, the super blow-out function ensures accurate dispensing for even the lowest volumes.
Catalog Number
4662060
Unit Size
Each
Number of Channels
12
Volume (Metric)
10 to 100 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorYellow
Number of Channels12
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)10 to 100 μL
Increments0.2μL
Color CodeYellow
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
8
Volume (Metric)
1 to 10 μL
Compatible Tips
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorPink
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePink
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
8
Volume (Metric)
5 to 50 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorYellow
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)5 to 50 μL
Increments0.02μL
Color CodeYellow
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
8
Volume (Metric)
10 to 100 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorYellow
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)10 to 100 μL
Increments0.2μL
Color CodeYellow
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
8
Volume (Metric)
30 to 300 μL
Compatible Tips
Finntip 300, Filter 300, Flex 300, Flex Filter 300
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorOrange
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)30 to 300 μL
Increments1μL
Color CodeOrange
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
12
Volume (Metric)
1 to 10 μL
Compatible Tips
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorPink
Number of Channels12
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePink
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
12
Volume (Metric)
5 to 50 μL
Compatible Tips
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorYellow
Number of Channels12
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeYellow
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
12
Volume (Metric)
30 to 300 μL
Compatible Tips
Finntip 300, Filter 300, Flex 300, Flex Filter 300
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorOrange
Number of Channels12
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)30 to 300 μL
Increments1μL
Color CodeOrange
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
16
Volume (Metric)
1 to 10 μL
Compatible Tips
Finntip 20 Micro, Filter 20 Micro
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 20 Micro, Filter 20 Micro
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorPurple
Number of Channels16
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePurple
Unit SizeEach
Catalog Number
Unit Size
Each
Number of Channels
16
Volume (Metric)
5 to 50 μL
Compatible Tips
Finntip 50 Micro, Filter 50 Micro
Price (USD)
Availability
-
Full specifications
Quantity
Compatible TipsFinntip 50 Micro, Filter 50 Micro
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorTurquoise Button
Number of Channels16
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeTurquoise
Unit SizeEach
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeNumber of ChannelsVolume (Metric)Compatible TipsPrice (USD)AvailabilityQuantity
4662060Full specifications
Each1210 to 100 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorYellow
Number of Channels12
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)10 to 100 μL
Increments0.2μL
Color CodeYellow
Unit SizeEach
4662000Full specifications
Each81 to 10 μLFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
-
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorPink
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePink
Unit SizeEach
4662010Full specifications
Each85 to 50 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorYellow
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)5 to 50 μL
Increments0.02μL
Color CodeYellow
Unit SizeEach
4662020Full specifications
Each810 to 100 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorYellow
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)10 to 100 μL
Increments0.2μL
Color CodeYellow
Unit SizeEach
4662030Full specifications
Each830 to 300 μLFinntip 300, Filter 300, Flex 300, Flex Filter 300
-
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorOrange
Number of Channels8
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)30 to 300 μL
Increments1μL
Color CodeOrange
Unit SizeEach
4662040Full specifications
Each121 to 10 μLFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
-
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorPink
Number of Channels12
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePink
Unit SizeEach
4662050Full specifications
Each125 to 50 μLFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
-
Compatible TipsFinntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorYellow
Number of Channels12
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeYellow
Unit SizeEach
4662070Full specifications
Each1230 to 300 μLFinntip 300, Filter 300, Flex 300, Flex Filter 300
-
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorOrange
Number of Channels12
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)30 to 300 μL
Increments1μL
Color CodeOrange
Unit SizeEach
4662080Full specifications
Each161 to 10 μLFinntip 20 Micro, Filter 20 Micro
-
Compatible TipsFinntip 20 Micro, Filter 20 Micro
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorPurple
Number of Channels16
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)1 to 10 μL
Increments0.02μL
Color CodePurple
Unit SizeEach
4662090Full specifications
Each165 to 50 μLFinntip 50 Micro, Filter 50 Micro
-
Compatible TipsFinntip 50 Micro, Filter 50 Micro
DescriptionFinnpipette™ F2
AutoclavableAutoclavable
ColorTurquoise Button
Number of Channels16
Product LineFinnpipette™ F2
TypePipette
Volume (Metric)5 to 50 μL
Increments0.1μL
Color CodeTurquoise
Unit SizeEach
Showing 10 of 10
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.