Thermo Scientific™
Heraguard™ ECO Clean Bench Accessories
Catalog number: 50073943
Have Questions?
Thermo Scientific™
Heraguard™ ECO Clean Bench Accessories
Catalog number: 50073943
Have Questions?
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
Catalog Number
50073943
Unit Size
Each
Product Type
General Accessories
Description
Hanging Bar 700 mm with set of 6 Hooks
For Use With (Equipment)
Heraguard ECO 0.9
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 0.9
DescriptionHanging Bar 700 mm with set of 6 Hooks
Product TypeGeneral Accessories
Configuration OptionsField Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
Media Valves
Description
Combustible gas G3/8 connection (yellow) with long neck for installation inside walls
For Use With (Equipment)
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8
DescriptionCombustible gas G3/8 connection (yellow) with long neck for installation inside walls
Product TypeMedia Valves
Configuration OptionsField Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
Media Valves
Description
Water tap G3/8 connection (Black) with long neck for installation in side walls
For Use With (Equipment)
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
DescriptionWater tap G3/8 connection (Black) with long neck for installation in side walls
Product TypeMedia Valves
Configuration OptionsField Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
Wind Screen Protection Kit
Description
Windscreen protection kit
For Use With (Equipment)
Heraguard ECO 0.9
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 0.9
DescriptionWindscreen protection kit
Product TypeWind Screen Protection Kit
Configuration OptionsFactory Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
Work Surface Wraps
Description
Stainless Steel Work Surface
For Use With (Equipment)
Heraguard ECO 0.9
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 0.9
Depth (Metric)580 mm
DescriptionStainless Steel Work Surface
Width (Metric)900 mm
Product TypeWork Surface Wraps
Configuration OptionsFactory Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
General Accessories
Description
Hanging Bar 1000 mm with Set of 10 Hooks
For Use With (Equipment)
Heraguard ECO 1.2, 1.5,1.8, 1.8/95
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 1.2, 1.5,1.8, 1.8/95
DescriptionHanging Bar 1000 mm with Set of 10 Hooks
Product TypeGeneral Accessories
Configuration OptionsField Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
Cover
Description
UV-resistant Night Cover
For Use With (Equipment)
Heraguard ECO 1.5
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 1.5
DescriptionUV-resistant Night Cover
Product TypeCover
Configuration OptionsField Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
Solenoid Valves
Description
Solenoid valve for use with tap for flammable gas cylinders G3/8 connection
For Use With (Equipment)
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
DescriptionSolenoid valve for use with tap for flammable gas cylinders G3/8 connection
Product TypeSolenoid Valves
Configuration OptionsField Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
UV Options
Description
Retrofit Kit UV-Radiator
For Use With (Equipment)
Heraguard ECO 1.5
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 1.5
DescriptionRetrofit Kit UV-Radiator
Product TypeUV Options
Configuration OptionsField Installed
Unit SizeEach
Catalog Number
Unit Size
Each
Product Type
Media Valves
Description
Lead through for hose connection for side wall use. Suitable for 10 – 13 mm inner diameter hose
For Use With (Equipment)
Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
Price (USD)
Full specifications
For Use With (Equipment)Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
DescriptionLead through for hose connection for side wall use. Suitable for 10 – 13 mm inner diameter hose
Product TypeMedia Valves
Configuration OptionsField Installed
Unit SizeEach
Showing 10 of 31Show more
Catalog NumberSpecificationsUnit SizeProduct TypeDescriptionFor Use With (Equipment)Price (USD)
50073943Full specifications
EachGeneral AccessoriesHanging Bar 700 mm with set of 6 HooksHeraguard ECO 0.9Request A Quote
For Use With (Equipment)Heraguard ECO 0.9
DescriptionHanging Bar 700 mm with set of 6 Hooks
Product TypeGeneral Accessories
Configuration OptionsField Installed
Unit SizeEach
50046015Full specifications
EachMedia ValvesCombustible gas G3/8 connection (yellow) with long neck for installation inside wallsHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8Request A Quote
For Use With (Equipment)Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8
DescriptionCombustible gas G3/8 connection (yellow) with long neck for installation inside walls
Product TypeMedia Valves
Configuration OptionsField Installed
Unit SizeEach
50044679Full specifications
EachMedia ValvesWater tap G3/8 connection (Black) with long neck for installation in side wallsHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95Request A Quote
For Use With (Equipment)Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
DescriptionWater tap G3/8 connection (Black) with long neck for installation in side walls
Product TypeMedia Valves
Configuration OptionsField Installed
Unit SizeEach
50052208Full specifications
EachWind Screen Protection KitWindscreen protection kitHeraguard ECO 0.9Request A Quote
For Use With (Equipment)Heraguard ECO 0.9
DescriptionWindscreen protection kit
Product TypeWind Screen Protection Kit
Configuration OptionsFactory Installed
Unit SizeEach
51901074Full specifications
EachWork Surface WrapsStainless Steel Work SurfaceHeraguard ECO 0.9Request A Quote
For Use With (Equipment)Heraguard ECO 0.9
Depth (Metric)580 mm
DescriptionStainless Steel Work Surface
Width (Metric)900 mm
Product TypeWork Surface Wraps
Configuration OptionsFactory Installed
Unit SizeEach
50073944Full specifications
EachGeneral AccessoriesHanging Bar 1000 mm with Set of 10 HooksHeraguard ECO 1.2, 1.5,1.8, 1.8/95Request A Quote
For Use With (Equipment)Heraguard ECO 1.2, 1.5,1.8, 1.8/95
DescriptionHanging Bar 1000 mm with Set of 10 Hooks
Product TypeGeneral Accessories
Configuration OptionsField Installed
Unit SizeEach
50139418Full specifications
EachCoverUV-resistant Night CoverHeraguard ECO 1.5Request A Quote
For Use With (Equipment)Heraguard ECO 1.5
DescriptionUV-resistant Night Cover
Product TypeCover
Configuration OptionsField Installed
Unit SizeEach
50074905Full specifications
EachSolenoid ValvesSolenoid valve for use with tap for flammable gas cylinders G3/8 connectionHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95Request A Quote
For Use With (Equipment)Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
DescriptionSolenoid valve for use with tap for flammable gas cylinders G3/8 connection
Product TypeSolenoid Valves
Configuration OptionsField Installed
Unit SizeEach
50140094Full specifications
EachUV OptionsRetrofit Kit UV-RadiatorHeraguard ECO 1.5Request A Quote
For Use With (Equipment)Heraguard ECO 1.5
DescriptionRetrofit Kit UV-Radiator
Product TypeUV Options
Configuration OptionsField Installed
Unit SizeEach
50059017Full specifications
EachMedia ValvesLead through for hose connection for side wall use. Suitable for 10 – 13 mm inner diameter hoseHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95Request A Quote
For Use With (Equipment)Heraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
DescriptionLead through for hose connection for side wall use. Suitable for 10 – 13 mm inner diameter hose
Product TypeMedia Valves
Configuration OptionsField Installed
Unit SizeEach
Showing 10 of 31Show more
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.