| 5400920 | Full specifications | Each | KIngFisher Apex with 96 Combi head | Request A Quote |
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | Suitable Downstream Analysis | PCR/qPCR, Sequencing, Mass Spec | Volume (Metric) Processing | KF 96 plate 15 to 200 μL, 96 DW plate 15 to 1000 μL, 96 storage tube 30 to 200 μL | Plates per Deck | 8 | Format | 96-well | Capacity | 96 Samples/Run | Description | KIngFisher Apex with 96 Combi head | Collection Efficiency of Particles | >98% with 2.8 μm particle size | Cooling Temperature | Down to +4°C | Display | Touch screen (10 in.) | For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | Heating Temperature | +4°C above ambient temperature up to +100°C | Magnetic Rods | 96 | Particle Size (Metric) | ca. >1 μm | Samples per Run | 96 | Type of Magnet Head | 96 in one frame, interchangeable | UV Exposure Time | Max 23 hr. 59 min. | UV Lamp | 2 pcs | Plate Type | 96 KF, 96 DW, 96 storeage tube | Depth (English) | 22 in. | Depth (Metric) | 56 cm | Height (English) | 18 in. | Height (Metric) | 46 cm | Weight (English) | 123 lb. | Weight (Metric) | 56 kg | Width (English) | 31 in. | Width (Metric) | 79 cm | Unit Size | Each |
|
| 5400910 | Full specifications | Each | KingFisher Apex with 96 PCR head | Request A Quote |
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | Suitable Downstream Analysis | PCR/qPCR, Sequencing, Mass Spec | Volume (Metric) Processing | 10 to 80 μL | Plates per Deck | 8 | Capacity | 96 Samples | Description | KingFisher Apex with 96 PCR head | Collection Efficiency of Particles | >98% with 2.8 μm particle size | Cooling Temperature | Down to +40°C | Display | Touch screen (10 in.) | For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | Heating Temperature | +4°C above ambient temperature up to +100°C | Magnetic Rods | 96 | Particle Size (Metric) | ca. >1 μm | Samples per Run | 96 | Type of Magnet Head | 96 in one frame, interchangeable | UV Exposure Time | Up to 23 hr. 59 min. | UV Lamp | 2 pcs | Plate Type | 96 PCR plate (semi/skirted) | Depth (English) | 22 in. | Depth (Metric) | 56 cm | Height (English) | 18 in. | Height (Metric) | 46 cm | Weight (English) | 123 lb. | Weight (Metric) | 56 kg | Width (English) | 31 in. | Width (Metric) | 79 cm | Unit Size | Each |
|
| A46941 | Full specifications | System + SmartStart + 1 yr ext warranty | KingFisher Apex with 96 PCR head, Extended Warranty Package | Request A Quote |
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | Suitable Downstream Analysis | PCR/qPCR, Sequencing, Mass Spec | Volume (Metric) Processing | 10 to 80 μL | Plates per Deck | 8 | Capacity | 96 Samples | Description | KingFisher Apex with 96 PCR head, Extended Warranty Package | Collection Efficiency of Particles | >98% with 2.8 μm particle size | Cooling Temperature | Down to +4°C | Display | Touch screen (10 in.) | For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | Heating Temperature | +4°C above ambient temperature up to +100°C | Magnetic Rods | 96 | Particle Size (Metric) | ca. >1 μm | Samples per Run | 96 | Type of Magnet Head | 96 in one frame, interchangeable | UV Exposure Time | Up to 23 hr. 59 min. | UV Lamp | 2 Pc. | Plate Type | 96 PCR plate (semi/skirted) | Depth (English) | 22 in. | Depth (Metric) | 56 cm | Height (English) | 18 in. | Height (Metric) | 46 cm | Weight (English) | 123 lb. | Weight (Metric) | 56 kg | Width (English) | 31 in. | Width (Metric) | 79 cm | Customer Education | 1 day at customer site | Total Service Coverage | 1 year manufacturer warranty and 1 year of field service with PM | Unit Size | System + SmartStart + 1 yr ext warranty |
|
| A46943 | Full specifications | System + SmartStart + 1 yr ext warranty | KingFisher Apex with 96 Combi head, Extended Warranty Package | Request A Quote |
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | Volume (Metric) Processing | KF 96 plate 15 to 200 μL, 96 DW plate 15 to 1000 μL, 96 storage tube 30 to 200 μL | Plates per Deck | 8 | Format | 96-well | Capacity | 96 Samples/Run | Description | KingFisher Apex with 96 Combi head, Extended Warranty Package | Collection Efficiency of Particles | >98% with 2.8 μm particle size | Cooling Temperature | Down to +4°C | For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | Heating Temperature | +4°C above ambient temperature up to +100°C | Magnetic Rods | 96 | Particle Size (Metric) | ca. >1 μm | Samples per Run | 96 | Type of Magnet Head | 96 in one frame, interchangeable | UV Exposure Time | Max 23 hr. 59 min. | UV Lamp | 2 Pc. | Plate Type | 96 KF, 96 DW, 96 storage tube | Depth (English) | 22 in. | Depth (Metric) | 56 cm | Height (English) | 18 in. | Height (Metric) | 46 cm | Weight (English) | 123 lb. | Weight (Metric) | 56 kg | Width (English) | 31 in. | Width (Metric) | 79 cm | Customer Education | 1 day at customer site | Total Service Coverage | 1 year manufacturer warranty and 1 year of field service with PM | Unit Size | System + SmartStart + 1 yr ext warranty |
|
| 5400930 | Full specifications | Each | KingFisher Apex with 96 DW head | Request A Quote |
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | Suitable Downstream Analysis | PCR/qPCR, Sequencing, Mass Spec | Volume (Metric) Processing | 96 KF plate 50 to 150 μL, 96 DW plate 50 to 1000 μL | Plates per Deck | 8 | Format | 96-well | Capacity | 96 Samples | Description | KingFisher Apex with 96 DW head | Collection Efficiency of Particles | >98% with 2.8 μm particle size | Cooling Temperature | Down to +4°C | Display | Touch screen (10 in.) | For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | Heating Temperature | +4°C above ambient temperature up to +100°C | Magnetic Rods | 96 in one frame, interchangeable | Particle Size (Metric) | ca. >1 μm | Samples per Run | 96 | Type of Magnet Head | 96 in one frame, interchangeable | UV Exposure Time | Max 23 hr. 59 min. | UV Lamp | 2 pcs | Plate Type | 96 KF, 96 DW | Depth (English) | 22 in. | Depth (Metric) | 56 cm | Height (English) | 18 in. | Height (Metric) | 46 cm | Weight (English) | 123 lb. | Weight (Metric) | 56 kg | Width (English) | 31 in. | Width (Metric) | 79 cm | Unit Size | Each |
|
| A46944 | Full specifications | System + SmartStart + 1 yr ext warranty | KingFisher Apex with 96 DW head, Extended Warranty Package | Request A Quote |
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | Suitable Downstream Analysis | PCR/qPCR, Sequencing, Mass Spec | Volume (Metric) Processing | 96 KF plate 50 to 150 μL, 96 DW plate 50 to 1000 μL | Plates per Deck | 8 | Format | 96-well | Capacity | 96 Samples | Description | KingFisher Apex with 96 DW head, Extended Warranty Package | Collection Efficiency of Particles | >98% with 2.8 μm particle size | Cooling Temperature | Down to +4°C | Display | Touch screen (10 in.) | For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | Heating Temperature | +4°C above ambient temperature up to +100°C | Magnetic Rods | 96 in one frame, interchangeable | Particle Size (Metric) | ca. >1 μm | Samples per Run | 96 | Type of Magnet Head | 96 in one frame, interchangeable | UV Exposure Time | Max 23 hr. 59 min. | UV Lamp | 2 Pc. | Plate Type | 96 KF, 96 DW | Depth (English) | 22 in. | Depth (Metric) | 56 cm | Height (English) | 18 in. | Height (Metric) | 46 cm | Weight (English) | 123 lb. | Weight (Metric) | 56 kg | Width (English) | 31 in. | Width (Metric) | 79 cm | Customer Education | 1 day at customer site | Total Service Coverage | 1 year manufacturer warranty and 1 year of field service with PM | Unit Size | System + SmartStart + 1 yr ext warranty |
|
| 5400940 | Full specifications | Each | KingFisher Apex with 24 Combi head | Request A Quote |
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | Suitable Downstream Analysis | PCR/qPCR, Sequencing, Mass Spec | Volume (Metric) Processing | 24 DW plate 30 to 5000 μL, 24 storage tube 200 to 1000 μL | Plates per Deck | 8 | Format | 24-well | Capacity | 24 Samples | Description | KingFisher Apex with 24 Combi head | Collection Efficiency of Particles | >98% with 2.8 μm particle size | Cooling Temperature | Down to +4°C | Display | Touch screen (10 in.) | For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | Heating Temperature | +4°C above ambient temperature up to +100°C | Magnetic Rods | 24 in one frame, interchangeable | Particle Size (Metric) | ca. >1 μm | Samples per Run | 24 | Type of Magnet Head | 24 | UV Exposure Time | Max 23 hr. 59 min. | UV Lamp | 2 pcs | Plate Type | 24 DW, 24 storage tube | Depth (English) | 22 in. | Depth (Metric) | 56 cm | Height (English) | 18 in. | Height (Metric) | 46 cm | Weight (English) | 123 lb. | Weight (Metric) | 56 kg | Width (English) | 31 in. | Width (Metric) | 79 cm | Unit Size | Each |
|
| A46942 | Full specifications | System + SmartStart + 1 yr ext warranty | KingFisher Apex with 24 Combi Head, Extended Warranty Package | Request A Quote |
Sample Type | Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate | Suitable Downstream Analysis | PCR/qPCR, Sequencing, Mass Spec | Volume (Metric) Processing | 24 DW plate 30 to 5000 μL, 24 storage tube 200 to 1000 μL | Plates per Deck | 8 | Format | 24-well | Capacity | 24 Samples | Description | KingFisher Apex with 24 Combi Head, Extended Warranty Package | Collection Efficiency of Particles | >98% with 2.8 μm particle size | Cooling Temperature | Down to +4°C | Display | Touch screen (10 in.) | For Use With (Application) | Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics | Heating Temperature | +4°C above ambient temperature up to +100°C | Magnetic Rods | 24 in one frame, interchangeable | Particle Size (Metric) | ca. >1 μm | Samples per Run | 24 | Type of Magnet Head | 24 | UV Exposure Time | Max 23 hr. 59 min. | UV Lamp | 2 Pc. | Plate Type | 24 DW, 24 storage tube | Depth (English) | 22 in. | Depth (Metric) | 56 cm | Height (English) | 18 in. | Height (Metric) | 46 cm | Weight (English) | 123 lb. | Weight (Metric) | 56 kg | Width (English) | 31 in. | Width (Metric) | 79 cm | Customer Education | 1 day at customer site | Total Service Coverage | 1 year manufacturer warranty and 1 year of field service with PM | Unit Size | System + SmartStart + 1 yr ext warranty |
|