Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ Disposable Filter Units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid Flow Filter Units and Bottle Tops are the best choice for the filtration of cell culture media, serum, additives and buffers. Some products on this page are ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions (see Full Specifications in the product table for item participation).
ACT Label | Yes |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
No. per Case | 12 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 250 mL |
Volume (Metric) Upper | 250 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
No. per Case | 12 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 50 mL |
Volume (Metric) Upper | 150 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.45 μm |
Sterility | Sterile |
No. per Case | 72 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 115 mL |
Volume (Metric) Upper | 115 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 72 |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.45 μm |
Sterility | Sterile |
No. per Case | 72 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 115 mL |
Volume (Metric) Upper | 115 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Pack of 12 |
ACT Label | Yes |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.1 μm |
Sterility | Sterile |
No. per Case | 12 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 150 mL |
Volume (Metric) Upper | 150 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
No. per Case | 72 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 115 mL |
Volume (Metric) Upper | 115 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 72 |
ACT Label | Yes |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.2 μm |
Sterility | Sterile |
No. per Case | 12 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 150 mL |
Volume (Metric) Upper | 150 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
ACT Label | Yes |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.45 μm |
Sterility | Sterile |
No. per Case | 12 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 150 mL |
Volume (Metric) Upper | 150 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
ACT Label | Yes |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.1 μm |
Sterility | Sterile |
No. per Case | 12 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 250 mL |
Volume (Metric) Upper | 250 mL |
Diameter (Metric) Membrane | 75 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
ACT Label | Yes |
Disposable | Single-use |
Material (Membrane) | PES |
Pore Size | 0.45 μm |
Sterility | Sterile |
No. per Case | 12 |
No. per Pack | 12 |
Type | Filter Unit |
Volume (Metric) Receiver | 250 mL |
Volume (Metric) Upper | 250 mL |
Diameter (Metric) Membrane | 50 mm |
Fit for Purpose | Sterile |
Unit Size | Case of 12 |
Catalog Number | Specifications | Unit Size | Material (Membrane) | Diameter (Metric) Membrane | Pore Size | Volume (Metric) Receiver | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
568-0020 | Case of 12 | PES | 50 mm | 0.2 μm | 250 mL | 121.60 Online price 152.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
564-0020 | Case of 12 | PES | 50 mm | 0.2 μm | 50 mL | 115.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
124-0045 | Case of 72 | PES | 50 mm | 0.45 μm | 115 mL | 612.90 Online price 681.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
124-0045PK | Pack of 12 | PES | 50 mm | 0.45 μm | 115 mL | 113.70 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
565-0010 | Case of 12 | PES | 50 mm | 0.1 μm | 150 mL | 96.40 Online price 116.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
524-0020 | Case of 72 | PES | 50 mm | 0.2 μm | 115 mL | 631.26 Online price 668.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
565-0020 | Case of 12 | PES | 50 mm | 0.2 μm | 150 mL | 94.40 Online price 118.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
165-0045 | Case of 12 | PES | 50 mm | 0.45 μm | 150 mL | 107.10 Online price 119.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
568-0010 | Case of 12 | PES | 75 mm | 0.1 μm | 250 mL | 126.11 Online price 150.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
168-0045 | Case of 12 | PES | 50 mm | 0.45 μm | 250 mL | 137.70 Online price 153.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|