Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin. These kits were designed to remove the challenges often associated with the process of protein cleanup by providing a simple and easy capture workflow.
Description | SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, SOLAμ Collection Plate, with Soluble Trypsin |
Capture | Streptavidin |
Cleavage | C-terminal side of lysine and arginine amino acids |
Enzyme | Trypsin |
Format | SMART Digest Resin, Bulk Magnetic |
For Use With (Application) | Immunocapture and Protein Digest Sample Preparation |
Product Line | SMART Digest |
Type | Immunocapture + Protein Digestion (separate) |
Includes | Bulk Magnetic SMART Digest Resin, SMART Digest Soluble Trypsin, Collection Plate, SOLAμ SPE Plate |
Unit Size | Each |
Description | SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, without Soluble Trypsin |
Capture | Streptavidin |
Cleavage | C-terminal side of lysine and arginine amino acids |
Enzyme | Trypsin |
Format | SMART Digest Resin, Bulk Magnetic |
For Use With (Application) | Immunocapture Sample Preparation |
Product Line | SMART Digest |
Type | Immunocapture |
Includes | Bulk Magnetic SMART Digest Resin |
Unit Size | Each |
Description | SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, with Soluble Trypsin |
Capture | Streptavidin |
Cleavage | C-terminal side of lysine and arginine amino acids |
Enzyme | Trypsin |
Format | SMART Digest Resin, Bulk Magnetic |
For Use With (Application) | Immunocapture and Protein Digest Sample Preparation |
Product Line | SMART Digest |
Type | Immunocapture + Protein Digestion (separate) |
Includes | Bulk Magnetic SMART Digest Resin, SMART Digest Soluble Trypsin |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
60119-103 | Each | SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, SOLAμ Collection Plate, with Soluble Trypsin | 1,849.11 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
60118-101 | Each | SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, without Soluble Trypsin | 902.05 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
60119-104 | Each | SMART Digest Bulk Fractionation Kit, Streptavidin, Magnetic, with Soluble Trypsin | 1,564.54 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The kit contains beads with an attached streptavidin moiety enabling a strong streptavidin-biotin bond with the appropriate biotinylated molecule with specific affinity for the target biomolecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.