1x1 image pixel for data collection
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Actual product may vary
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Thermo Scientific™
Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Use Thermo Scientific™ Manual Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
60180-DCR1313 mm Crimp VialsManual Decrimper
60180-CR1111 mm Crimp VialsManual Crimper
60180-CR1313 mm Crimp VialsManual Crimper
60180-CR2020 mm Crimp VialsManual Crimper
60180-CR88 mm Crimp VialsManual Crimper
60180-DCR1111 mm Crimp VialsManual Decrimper
60180-DCR2020 mm Crimp VialsManual Decrimper
Catalog number 60180-DCR13
Price (USD)
434.94
Each
Add to cart
For Use With (Equipment):
13 mm Crimp Vials
Type:
Manual Decrimper
Price (USD)
434.94
Each
Add to cart
Ask our AI about this Product
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
Specifications
For Use With (Equipment)13 mm Crimp Vials
MaterialStainless Steel/Aluminum
Product LineSureSTART Level 2
TypeManual Decrimper
Unit SizeEach
SureSTART Vials and Caps
Selector tools
Tools designed to simplify your search for the perfect SureSTART product for your workflow.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Videos
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection