Dichloromethane, for HPLC
Dichloromethane, for HPLC
Dichloromethane, for HPLC
Dichloromethane, for HPLC
Thermo Scientific Chemicals

Dichloromethane, for HPLC

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Catalog NumberQuantityPackaging
6100500404 LEcoSafPak™
Catalog number 610050040
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
-
Add to cart
Quantity:
4 L
Packaging:
EcoSafPak™
Request bulk or custom format
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.42 to 1.423 (25°C, 589 nm)
GC>=99.9 % Including preservative
Water=<0.02 %
Titrable acid=<0.0003 meq/g
Residue after evaporation=<2 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection