Informational:Regulatory requirements vary by country; products may not be available in your geographic area.
Thermo Scientific™

BOVIGAM™ TB Kit

Catalog number: 63326
Have Questions?
Thermo Scientific™

BOVIGAM™ TB Kit

Catalog number: 63326
Have Questions?
Catalog Number
63326
Unit Size
450 samples
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
150 samples
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
63326450 samples
--
63320150 samples
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The BOVIGAM test is a blood-based in vitro laboratory test for bovine tuberculosis. It is based on the detection of cell-mediated immune response to infection with Mycobacterium bovis in cattle, sheep, goats, buffalo, bison, and other bovidae. The BOVIGAM test involves two stages. In the first stage, blood samples are incubated overnight with antigens, such as tuberculin purified protein derivative (PPD) or the newly developed 'peptide cocktails', to stimulate lymphocytes to produce IFN-γ. In the second stage, IFN-γ in the plasma supernatants of each blood aliquot is determined using a sandwich ELISA. IFN-γ present in the sample binds to anti-bovine IFN-γ monoclonal antibodies on a solid support and is visualized with a second anti-IFN-γ antibody labeled with an enzyme that generates a color signal. Color development is proportional to the amount of bound IFN-γ.

The BOVIGAM test is officially approved by the World Organisation for Animal Health (WOAH, formerly known as OIE), indicating that it has successfully met the rigorous standards established by WOAH—a globally esteemed authority in animal health. This endorsement validates BOVIGAM, affirming its suitability and effectiveness for all situations requiring bovine tuberculosis (bTB) testing.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Specifications

Assay Sensitivity
See product documentation
Immunoassay Kit Format
Other Format
Product Line
BOVIGAM™, Prionics™
Protein Family
Other Proteins
Target Species
Bovine
Gene ID (Entrez)
Non-Gene
Gene Symbol
Non-Gene
Quantity
450 Samples
Target Kit Named
TB
Target Organism Class
Mycobacterium bovis
Target Species Validated
Bovine, Goat, Sheep

Contents & Storage

30 plates

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text