Invitrogen™

eBioscience™ Cell Proliferation Dye eFluor™ 670

Catalog number: 65-0840-90
Have Questions?
Invitrogen™

eBioscience™ Cell Proliferation Dye eFluor™ 670

Catalog number: 65-0840-90
Have Questions?
Catalog Number
65-0840-90
Unit Size
4 x 500 µg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
65-0840-904 x 500 µg
-
65-0840-85500 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Cell Proliferation Dye eFluor™ 670 is a red fluorescent dye that can be used to monitor individual cell divisions. This fluorescent dye binds to any cellular protein containing primary amines, and as cells divide, the dye is distributed equally between daughter cells that can be measured as successive halving of the fluorescence intensity of the dye. Up to 6 generations may be visualized. Cell Proliferation Dye eFluor™ 670 can also be used for long term tracking of labeled cells. Analysis using two-parameter plots may provide better resolution of each generation, especially between undivided cells and the first generation. Cells labeled with Cell Proliferation Dye eFluor™ 670 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers, such as the Foxp3 Transcription Factor Staining Buffer Set (cat. 00-5523) or the IC Fixation Buffer (cat. 00-8222) and Permeabilization Buffer (10X) (cat. 00-8333).

Cell Proliferation Dye eFluor™ 670 has a peak excitation of 647 nm and can be excited by the red (633 nm) laser line. It has a peak emission of 670 nm and can be detected with a 660/20 band pass filter (equivanet to APC, Alexa Fluor™ 647, or eFluor™ 660), making it compatible with applications that utilize GFP.

Cell Proliferation Dye eFluor™ 670 has a molecular weight of 792.6 and is supplied as a lyophilized powder. Each vial may be reconstituted to a stock concentration of 5 mM with 126 μL of anhydrous DMSO; once reconstituted it should be protected from light and stored at -20°C with dessicant. It is recommended to use the reconstituted dye within 6 months and to avoid freeze-thawing.

Reported Application
Flow Cytometric Analysis, Microscopy

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Red
Detection Method
Fluorescent
For Use With (Application)
Proliferation Assay
For Use With (Equipment)
Fluorescence Microscope, Flow Cytometer
Product Type
Dye
Emission
630/670
Form
Lyophilized powder
Product Line
eBioscience™
Quantity
4 x 500 μg

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text