Thermo Scientific™

Slide-A-Lyzer™ Dialysis Cassettes, 20K MWCO

Catalog number: 66005
Have Questions?
Thermo Scientific™

Slide-A-Lyzer™ Dialysis Cassettes, 20K MWCO

Catalog number: 66005
Have Questions?
Slide-A-Lyzer Dialysis Cassettes (20K MWCO) facilitate the simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules larger than 20,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange, and more complete sample recovery.
 
Catalog Number
66005
Unit Size
10 cassettes
Sample Volume (Metric)
0.5 mL
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 0.5mL
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO20 kDa
Product LineSlide-A-Lyzer™
Quantity10 Cassettes
Sample Volume (Metric)0.5 mL
Unit Size10 cassettes
Catalog Number
Unit Size
10 cassettes
Sample Volume (Metric)
3 mL
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 3mL
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO20 kDa
Product LineSlide-A-Lyzer™
Quantity10 Cassettes
Sample Volume (Metric)3 mL
Unit Size10 cassettes
Catalog Number
Unit Size
8 cassettes
Sample Volume (Metric)
12 mL
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 12mL
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO20 kDa
Product LineSlide-A-Lyzer™
Quantity8 Cassettes
Sample Volume (Metric)12 mL
Unit Size8 cassettes
Catalog Number
Unit Size
6 cassettes
Sample Volume (Metric)
30 mL
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 30mL
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO20 kDa
Product LineSlide-A-Lyzer™
Quantity6 Cassettes
Sample Volume (Metric)30 mL
Unit Size6 cassettes
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeSample Volume (Metric)Price (USD)AvailabilityQuantity
66005Full specifications
10 cassettes0.5 mL
-
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 0.5mL
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO20 kDa
Product LineSlide-A-Lyzer™
Quantity10 Cassettes
Sample Volume (Metric)0.5 mL
Unit Size10 cassettes
66003Full specifications
10 cassettes3 mL
-
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 3mL
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO20 kDa
Product LineSlide-A-Lyzer™
Quantity10 Cassettes
Sample Volume (Metric)3 mL
Unit Size10 cassettes
66012Full specifications
8 cassettes12 mL
-
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 12mL
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO20 kDa
Product LineSlide-A-Lyzer™
Quantity8 Cassettes
Sample Volume (Metric)12 mL
Unit Size8 cassettes
66030Full specifications
6 cassettes30 mL
-
DescriptionSlide-A-Lyzer Dialysis Cassettes, 20K MWCO, 30mL
Purification TargetBuffer Exchange, Protein
MembraneCellulose Membrane, Dialysis
FormatCassette
MWCO20 kDa
Product LineSlide-A-Lyzer™
Quantity6 Cassettes
Sample Volume (Metric)30 mL
Unit Size6 cassettes
Showing 4 of 4
Slide-A-Lyzer Dialysis Cassettes (20K MWCO) facilitate the simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules larger than 20,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange and more complete sample recovery. Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Four cassette sizes—select the cassette that best suits your sample volume
Color-coded—purple cassette frame specifies cassettes with the 20,000 MW pore size membrane

Membrane specifications:
Composition: regenerated cellulose synthesized by the Viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: none
Sulfur content: 0.04%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

For Research Use Only. Not for use in diagnostic procedures.

Contents & Storage

Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text