Thermo Scientific™
nanoViper™ Capillary Kits for Low-Flow UHPLC Systems
Catalog number: 6720.0395
Have Questions?
Thermo Scientific™
nanoViper™ Capillary Kits for Low-Flow UHPLC Systems
Catalog number: 6720.0395
Have Questions?
Avoid data-destructive dead volumes when connecting your low-flow UHPLC System. Ensure easy connections with the Thermo Scientific™ Standard Application Kit for Thermo Scientific™ Vanquish™ Neo UHPLC System for multiple nano-, capillary-, and micro-flow LC-MS workflows. Use the Thermo Scientific™ nanoViper™ MS Connection Kits for Thermo Scientific™ EASY-Spray™ Columns for ideal column connections to a Thermo Scientific™ UltiMate™ 3000 RSLCnano System. Use the dedicated MS connection kit for all other connections of the UltiMate 3000 RSLCnano system to an MS inlet source. A single kit contains all the connectors and capillaries you need for tight and consistent connections.
See Viper and nanoViper resources
Catalog Number
6720.0395
Unit Size
Each
Description
nanoViper™ EASY-Spray Connection Kit
For Use With (Equipment)
Dionex UltiMate 3000 RSLC Nano System
Price (USD)
Full specifications
DescriptionnanoViper™ EASY-Spray Connection Kit
For Use With (Application)LC-MS connection
For Use With (Equipment)Dionex UltiMate 3000 RSLC Nano System
MaterialShielded Fused Silica
Max. Pressure1200 bar
Product LinenanoViper
TypeAdvanced Capillary Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
nanoViper™ MS Connection Kit for Low-Flow UHPLC Systems
For Use With (Equipment)
UltiMate 3000 RSLCnano System and EASY-nLC 1200 System
Price (USD)
Full specifications
DescriptionnanoViper™ MS Connection Kit for Low-Flow UHPLC Systems
For Use With (Application)LC-MS connection
For Use With (Equipment)UltiMate 3000 RSLCnano System and EASY-nLC 1200 System
MaterialShielded Fused Silica
Max. Pressure1200 bar
Product LinenanoViper
TypeAdvanced Capillary Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Standard Application Kit for Vanquish Neo UHPLC Systems
For Use With (Equipment)
Vanquish Neo UHPLC System
Price (USD)
Full specifications
DescriptionStandard Application Kit for Vanquish Neo UHPLC Systems
For Use With (Application)Direct Injection, Trap-And-Elute, Heated Trap-And-Elute Workflows
For Use With (Equipment)Vanquish Neo UHPLC System
MaterialShielded Fused Silica
Max. Pressure1500 bar
Product LinenanoViper
TypeAdvanced Capillary Kit
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionFor Use With (Equipment)Price (USD)
6720.0395Full specifications
EachnanoViper™ EASY-Spray Connection KitDionex UltiMate 3000 RSLC Nano SystemRequest A Quote
DescriptionnanoViper™ EASY-Spray Connection Kit
For Use With (Application)LC-MS connection
For Use With (Equipment)Dionex UltiMate 3000 RSLC Nano System
MaterialShielded Fused Silica
Max. Pressure1200 bar
Product LinenanoViper
TypeAdvanced Capillary Kit
Unit SizeEach
6720.0345Full specifications
EachnanoViper™ MS Connection Kit for Low-Flow UHPLC SystemsUltiMate 3000 RSLCnano System and EASY-nLC 1200 SystemRequest A Quote
DescriptionnanoViper™ MS Connection Kit for Low-Flow UHPLC Systems
For Use With (Application)LC-MS connection
For Use With (Equipment)UltiMate 3000 RSLCnano System and EASY-nLC 1200 System
MaterialShielded Fused Silica
Max. Pressure1200 bar
Product LinenanoViper
TypeAdvanced Capillary Kit
Unit SizeEach
6250.1010Full specifications
EachStandard Application Kit for Vanquish Neo UHPLC SystemsVanquish Neo UHPLC SystemRequest A Quote
DescriptionStandard Application Kit for Vanquish Neo UHPLC Systems
For Use With (Application)Direct Injection, Trap-And-Elute, Heated Trap-And-Elute Workflows
For Use With (Equipment)Vanquish Neo UHPLC System
MaterialShielded Fused Silica
Max. Pressure1500 bar
Product LinenanoViper
TypeAdvanced Capillary Kit
Unit SizeEach
Showing 3 of 3
Experience the easy installation and robust performance of LC-MS connections with the nanoViper Fitting System kits for low-flow UHPLC Systems. Our nanoViper Fittings are designed to seal at the tip of the tubing to virtually eliminate dead volume and ensure easy connections in your low-flow UHPLC system.
Standard applications kits for the Vanquish Neo UHPLC system enable direct injection, trap-and-elute, and heated trap-and-elute LC-MS workflows. All kits facilitate dispersion-optimized MS front-end installation of the Thermo Scientific low-flow LC systems, including UltiMate 3000 RSLCnano systems, EASY-nLC 1200 systems, and Vanquish Neo UHPLC systems.
These kits feature:
  • Tailored kits for easy installation of MS front ends
  • Virtually zero-dead volume connections
  • Easy-to-install, reusable capillaries
  • Low carryover and simplified method transfer
  • Reliable and reproducible LC connections