Search Thermo Fisher Scientific
Secure your LC column in the Thermo Scientific™ Vanquish™ Column Compartment module using our column holders (for Vanquish Compartments H and C) and column clips (for Vanquish Column Compartment N).
For Use With (Equipment) | Vanquish Column Compartment H, Vanquish Column Compartment C |
Type | Column Holder |
Unit Size | Pack of 6 |
Type | Column Clip kit |
For Use With (Equipment) | Vanquish Column Compartment N |
Unit Size | Pack of 6 |
Catalog Number | Specifications | Unit Size | Type | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
6732.1904 | Pack of 6 | Column Holder | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
6722.0290 | Pack of 6 | Column Clip kit | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|