BetaSil™ C18 Javelin™ Guard Columns
Catalog number: 70105-012106
Have Questions?
BetaSil™ C18 Javelin™ Guard Columns
Catalog number: 70105-012106
Have Questions?
Offer convenient, easy to use protection for analytical HPLC columns
Catalog Number
70105-012106
Unit Size
Pack of 4
Diameter (Metric) Inner
2.1 mm
Length (Metric)
10 mm
Price (USD)
Full specifications
Particle Size5 μm
Length (Metric)10 mm
Diameter (Metric) Inner2.1 mm
Stationary PhaseC18
Product LineBetaSil
Unit SizePack of 4
Catalog Number
Unit Size
Pack of 4
Diameter (Metric) Inner
2.1 mm
Length (Metric)
20 mm
Price (USD)
Full specifications
Particle Size3 μm
Length (Metric)20 mm
Diameter (Metric) Inner2.1 mm
Stationary PhaseC18
Product LineBetaSil
Unit SizePack of 4
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDiameter (Metric) InnerLength (Metric)Price (USD)
70105-012106Full specifications
Pack of 42.1 mm10 mmRequest A Quote
Particle Size5 μm
Length (Metric)10 mm
Diameter (Metric) Inner2.1 mm
Stationary PhaseC18
Product LineBetaSil
Unit SizePack of 4
70105-022106Full specifications
Pack of 42.1 mm20 mmRequest A Quote
Particle Size3 μm
Length (Metric)20 mm
Diameter (Metric) Inner2.1 mm
Stationary PhaseC18
Product LineBetaSil
Unit SizePack of 4
Showing 2 of 2
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Betasil C18 Solid Phase
  • Ligand on 100Å silica
  • High surface area with high bonded phase coverage