Search Thermo Fisher Scientific
This is a replacement part for Nalgene™ 4300- and 4301- Separatory Funnels.
Perform a variety of lab duties with chemically inert, translucent and autoclavable Thermo Scientific™ Replacement Stopcock Assembly for Nalgene™ Polypropylene Separatory Funnels with Closure. Closure has excellent chemical resistance and requires no lubrication.
Material | TFE |
Description | Replacement Teflon Stopcock Assembly |
For Use With (Equipment) | Nalgene™ 4300- and 4301- Separatory Funnels |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | For Use With (Equipment) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
714301-1025 | Each | Nalgene™ 4300- and 4301- Separatory Funnels | 86.40 Online price 108.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Includes: