Thermo Scientific™
H-Flex 1 Swinging Bucket Rotor and Accessories
Catalog number: 75003309
Have Questions?
Thermo Scientific™
H-Flex 1 Swinging Bucket Rotor and Accessories
Catalog number: 75003309
Have Questions?
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
Catalog Number
75003309
also known as  75003309
Unit Size
Each
Description
Thermo Scientific H-Flex 1 Rotor Replacement O-Rings, set/2
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionThermo Scientific H-Flex 1 Rotor Replacement O-Rings, set/2
MaterialEPDM
TypeSwinging Bucket Rotor
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
BiocontainmentYes
AutoclavableNot Autoclavable
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific H-Flex 1 Rotor Cross
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionThermo Scientific H-Flex 1 Rotor Cross
MaterialCast/Machined Steel with Slide Coating
TypeSwinging Bucket Rotor
CapacitySee Individual Adapter Capacities
Max. Speed4297
Radius (Metric)Rmax 174 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
Angle90°
Certifications/ComplianceBiocontainment Certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes, when used with biocontainment lids
AutoclavableAutoclavable
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific H-Flex 1 Rotor Buckets, set/2
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionThermo Scientific H-Flex 1 Rotor Buckets, set/2
MaterialForged/Machined Aluminum with Anodized Coating
TypeSwinging Bucket Rotor
CapacitySee Individual Adapter Capacities
Max. Speed4297
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
Angle90°
Certifications/ComplianceBiocontainment Certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes, when used with biocontainment lids
AutoclavableAutoclavable
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific H-Flex 1 Rotor Biocontainment Lids, set/2
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionThermo Scientific H-Flex 1 Rotor Biocontainment Lids, set/2
MaterialMolded Polyetherimide
TypeSwinging Bucket Rotor
CapacitySee Individual Adapter Capacities
Max. Speed4297
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
Certifications/ComplianceBiocontainment Certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes, when used with buckets
AutoclavableAutoclavable (without o-ring)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific H-Flex 1 Rotor Microplate Adapter, set/2
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionThermo Scientific H-Flex 1 Rotor Microplate Adapter, set/2
MaterialMolded Polyamide Nylone
TypeSwinging Bucket Rotor
Capacity5 Microplates or 2 Deepwell Plates Each Adapter
Max. Speed4297
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
BiocontainmentYes
AutoclavableAutoclavable
Unit SizeEach
Showing 5 of 5
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
75003309
also known as  75003309
Full specifications
EachThermo Scientific H-Flex 1 Rotor Replacement O-Rings, set/2Request A Quote--
DescriptionThermo Scientific H-Flex 1 Rotor Replacement O-Rings, set/2
MaterialEPDM
TypeSwinging Bucket Rotor
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
BiocontainmentYes
AutoclavableNot Autoclavable
Unit SizeEach
75003300Full specifications
EachThermo Scientific H-Flex 1 Rotor Cross
-
DescriptionThermo Scientific H-Flex 1 Rotor Cross
MaterialCast/Machined Steel with Slide Coating
TypeSwinging Bucket Rotor
CapacitySee Individual Adapter Capacities
Max. Speed4297
Radius (Metric)Rmax 174 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
Angle90°
Certifications/ComplianceBiocontainment Certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes, when used with biocontainment lids
AutoclavableAutoclavable
Unit SizeEach
75003301Full specifications
EachThermo Scientific H-Flex 1 Rotor Buckets, set/2
-
DescriptionThermo Scientific H-Flex 1 Rotor Buckets, set/2
MaterialForged/Machined Aluminum with Anodized Coating
TypeSwinging Bucket Rotor
CapacitySee Individual Adapter Capacities
Max. Speed4297
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
Angle90°
Certifications/ComplianceBiocontainment Certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes, when used with biocontainment lids
AutoclavableAutoclavable
Unit SizeEach
75003302Full specifications
EachThermo Scientific H-Flex 1 Rotor Biocontainment Lids, set/2
-
DescriptionThermo Scientific H-Flex 1 Rotor Biocontainment Lids, set/2
MaterialMolded Polyetherimide
TypeSwinging Bucket Rotor
CapacitySee Individual Adapter Capacities
Max. Speed4297
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
Certifications/ComplianceBiocontainment Certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes, when used with buckets
AutoclavableAutoclavable (without o-ring)
Unit SizeEach
75003307Full specifications
EachThermo Scientific H-Flex 1 Rotor Microplate Adapter, set/2
-
DescriptionThermo Scientific H-Flex 1 Rotor Microplate Adapter, set/2
MaterialMolded Polyamide Nylone
TypeSwinging Bucket Rotor
Capacity5 Microplates or 2 Deepwell Plates Each Adapter
Max. Speed4297
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
BiocontainmentYes
AutoclavableAutoclavable
Unit SizeEach
Showing 5 of 5
  • Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
  • Spins blood tubes, conical tubes and microplates without changing rotor or buckets
  • Get results faster without the need to change hardware
  • Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants