Thermo Scientific™

Halt™ Protease and Phosphatase Inhibitor Cocktail (100X)

Catalog number: 78444
Have Questions?
Thermo Scientific™

Halt™ Protease and Phosphatase Inhibitor Cocktail (100X)

Catalog number: 78444
Have Questions?
Catalog Number
78444
Unit Size
5 x 1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
24 x 100 µL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
784445 x 1 mL
-
7844224 x 100 µL
-
7844610 mL
-
784401 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Halt Protease and Phosphatase Inhibitor Cocktail (100X) provides the convenience of a single solution with full protein sample protection for cell and tissue lysates.

Features of Halt Protease and Phosphatase Inhibitor Cocktail (100X):

Complete protection—all-in-one cocktail contains both protease and phosphatase inhibitors
Multiple package sizes—choose from four convenient packages sizes to match nearly any scale
Convenient —the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats
Easy storage—aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails
Compatible—can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift
Validated—fully compatible with Thermo Scientific Pierce Cell Lysis Buffers

Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases. To promote the inhibition of metalloproteases, 0.5M EDTA is provided in a separate tube.

Related Products
Halt™ Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Halt Protease and Phosphatase Inhibitor Cocktail
Product Type
Protease and Phosphatase Inhibitor Cocktail
Form
Liquid
Product Line
Halt™
Buffer
Lysis Buffer
Reagent Type
Protease Inhibitor, Phosphatase Inhibitor
Inhibitors
Protease Inhibitor, Phosphatase Inhibitor
Quantity
5 x 1 mL

Contents & Storage

Upon receipt store at 4°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text