Thermo Scientific™
Insight™ Pro Software
Catalog number: 840-371000
Have Questions?
Thermo Scientific™
Insight™ Pro Software
Catalog number: 840-371000
Have Questions?
Streamline your routine, complex and advanced routines and workflows with Thermo Scientific™ Insight™ Pro Software. Prevent costly sampling and measurement errors with prompting, step-by-step workflows. Add Thermo Scientific™ Insight™ Pro Security Software to achieve compliance with current FDA 21 CFR Part 11 regulations.
When installation and operational qualifications (IQ/OQ) are required, Thermo Scientific™ Validator™ for Insight Pro Software provides all the documentation and reference materials needed to complete IQ/OQ for Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers.
Catalog Number
840-371000
Unit Size
Each
Type
Validator for Insight Pro - A4 Version
Price (USD)
Availability
-
Full specifications
Quantity
-
DescriptionFeatures: IQ/OQ procedures for spectrophotometers, accessories, and software, Easy to follow steps that reference the included uncertified filter set, High quality binder A4 paper, Integrated sign-off in the IQ streamlines the process, Test result sheets for OQ procedures saves paper and makes qualification easy Includes: Binder with pre-printed tabs and DVD with: IQ/OQ procedures for spectrophotometers, accessories, and software, Concepts of validation documentation, Design Qualification section detailing vendor practices, Best practice guide for Performance Verification, Preventive maintenance information and forms, Incident report sheets and template masters, Preprinted master copies of certain core documents are included, Documents not required by all users must be printed from the DVD, Spectrophotometer Test Kit with calibrated filters (840-285200) referenced in the qualification procedures.
TypeValidator for Insight Pro - A4 Version
For Use With (Equipment)For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Insight Pro Software
Price (USD)
Availability
-
Full specifications
Quantity
TypeInsight Pro Software
For Use With (Equipment)For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Insight Pro Security Software
Price (USD)
Availability
-
Full specifications
Quantity
Certifications/ComplianceSupports current 21 CFR Part 11 compliance
TypeInsight Pro Security Software
For Use With (Equipment)For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Validator for Insight Pro - US Version
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionFeatures: IQ/OQ procedures for spectrophotometers, accessories, and software, Easy to follow steps that reference the included uncertified filter set, High quality binder for 8.5 inch x 11 inch paper, Integrated sign-off in the IQ streamlines the process, Test result sheets for OQ procedures saves paper and makes qualification easyIncludes: Binder with pre-printed tabs and DVD with: IQ/OQ procedures for spectrophotometers, accessories, and software, Concepts of validation documentation, Design Qualification section detailing vendor practices, Best practice guide for Performance Verification, Preventive maintenance information and forms, Incident report sheets and template masters, Preprinted master copies of certain core documents are included, Documents not required by all users must be printed from the DVD, Spectrophotometer Test Kit with calibrated filters (840-285200) referenced in the qualification procedures.
TypeValidator for Insight Pro - US Version
For Use With (Equipment)For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeTypePrice (USD)AvailabilityQuantity
840-371000Full specifications
EachValidator for Insight Pro - A4 VersionRequest A Quote--
DescriptionFeatures: IQ/OQ procedures for spectrophotometers, accessories, and software, Easy to follow steps that reference the included uncertified filter set, High quality binder A4 paper, Integrated sign-off in the IQ streamlines the process, Test result sheets for OQ procedures saves paper and makes qualification easy Includes: Binder with pre-printed tabs and DVD with: IQ/OQ procedures for spectrophotometers, accessories, and software, Concepts of validation documentation, Design Qualification section detailing vendor practices, Best practice guide for Performance Verification, Preventive maintenance information and forms, Incident report sheets and template masters, Preprinted master copies of certain core documents are included, Documents not required by all users must be printed from the DVD, Spectrophotometer Test Kit with calibrated filters (840-285200) referenced in the qualification procedures.
TypeValidator for Insight Pro - A4 Version
For Use With (Equipment)For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Unit SizeEach
837-003900Full specifications
EachInsight Pro Software
-
TypeInsight Pro Software
For Use With (Equipment)For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Unit SizeEach
837-004000Full specifications
EachInsight Pro Security Software
-
Certifications/ComplianceSupports current 21 CFR Part 11 compliance
TypeInsight Pro Security Software
For Use With (Equipment)For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Unit SizeEach
840-370900Full specifications
EachValidator for Insight Pro - US Version
-
DescriptionFeatures: IQ/OQ procedures for spectrophotometers, accessories, and software, Easy to follow steps that reference the included uncertified filter set, High quality binder for 8.5 inch x 11 inch paper, Integrated sign-off in the IQ streamlines the process, Test result sheets for OQ procedures saves paper and makes qualification easyIncludes: Binder with pre-printed tabs and DVD with: IQ/OQ procedures for spectrophotometers, accessories, and software, Concepts of validation documentation, Design Qualification section detailing vendor practices, Best practice guide for Performance Verification, Preventive maintenance information and forms, Incident report sheets and template masters, Preprinted master copies of certain core documents are included, Documents not required by all users must be printed from the DVD, Spectrophotometer Test Kit with calibrated filters (840-285200) referenced in the qualification procedures.
TypeValidator for Insight Pro - US Version
For Use With (Equipment)For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Unit SizeEach
Showing 4 of 4
Make your lab time more productive with Insight Pro Software
  • Reduce training time - Easy to teach and learn, optimized user interface with prompting software defines clear step-by- step workflows
  • Prevent costly sampling and measurement errors - Create dedicated workflows for complex assay and transform multistep workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
  • Always current - Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
Streamlined QA/QC Analysis:
  • Automated result calculation options eliminate manual calculations and reduce user error.
  • Detailed graphical displays of calibration curves and sample run charts provide quick visual checks of system suitability and data trends.
  • Control limits and automated pass/fail interpretation provide immediate compliance results.
  • Scanning Quant includes ability to perform multiple calculations and extract parameters to create a complete analytical picture, improving method development experience.
Simplified Research Applications:
  • Multitude of data collection options and graphing features.
  • Pre-programmed peak analysis and convenient spectral processing tools aid in sample identification and quantitation for scanning measurements.
  • Multi-staged curve fitting and consecutive reaction modes provide comprehensive data fitting options for kinetics experiments.
  • Sophisticated math analysis functions, including derivatives and smoothing allow custom data display.
  • Merge workbooks feature allows quick and convenient comparison of data from multiple experiments.
Integrated Accessories:
  • Automated accessories allow complete control for superior performance with advanced sample handling requirements.
  • Calibration and alignment tools ensure accessories are performing optimally.
  • Supported temperature control accessories control and monitor reaction temperature in eight positions.
  • Programmable sipper, cell changer or autosampler accessory performs high-throughput activities with ease.
  • Hands-free performance verification and accessory calibration modules are designed for the SMART Sipper or Linear 8-Cell Changer accessory.
Comply with US FDA 21 CFR Part 11:
To ensure the security and integrity of your data and to help your lab comply with 21 CFR Part 11 or other regulations, add Insight Pro Security Software with which pairs Insight Pro Software with Thermo Scientific™ Security Suite Software.
  • Control user account access
  • Set and manage security policies to ensure proper data handling
  • Customize and enforce digital signatures to establish responsibility
  • Log and view security events
  • Electronic audit trails for a reliable electronic documentation
  • Database storage ensures to maintain electronic records for FDA submissions
Instrument Qualification:
Validator for Insight Pro Software includes:
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance
  • Installation and instrument certification