Thermo Scientific™

Brij™-35 Surfact-Amps™ Detergent Solution

Catalog number: 85117
Have Questions?
Thermo Scientific™

Brij™-35 Surfact-Amps™ Detergent Solution

Catalog number: 85117
Have Questions?
Catalog Number
85117
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
8511750 mL
-
283166 x 10 mL
-
85118500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Thermo Scientific™ formulation of Brij™ 35 is a stabilized 10% (w/v) solution of Brij-35 detergent for use in various protein methods. Brij™ 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications.

Properties of Brij 35 Detergent:

• Molecular Weight: 1225g
• Detergent Class: Nonionic
• Aggregation Number: 40
• Micelle Molecular Weight: 49,000g
• Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
• Cloud Point: >100°C
• Dialyzable: No

Features of this Thermo Scientific detergent solution:

Brij 35—a nonionic detergent for use in various protein methods
Accurate—10% detergent solution in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use

Specifications of this Thermo Scientific Brij 35, 10% Solution:

• Visual: Clear, colorless viscous liquid, free of foreign material.
• Concentration: Concentration: 27.0 to 30.0%

Related Products
Brij™-35 Surfact-Amps™ (10% w/v) Solution
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Brij-35 Surfact-Amps Detergent Solution
Product Type
Detergent
Form
Liquid
Product Line
Brij™, Surfact-Amps™
Reagent Type
Detergent Solution
Quantity
50 mL

Contents & Storage

Store at Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text