Thermo Scientific™

NP-40 Surfact-Amps™ Detergent Solution

Catalog number: 85125
Have Questions?
Thermo Scientific™

NP-40 Surfact-Amps™ Detergent Solution

Catalog number: 85125
Have Questions?
Catalog Number
85125
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
85125500 mL
-
8512450 mL
-
283246 x 10 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized as a 10% solution in your choice of either 10 mL glass ampules or two sizes of HDPE-plastic bottles.

Features of Surfact-Amps NP-40 Detergent Solution:

NP-40—a nonionic detergent for use in various protein methods
Accurate—precise 10% detergent solutions in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use
Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
Stable—packaged under inert nitrogen gas in HDPE bottles

This is an easy-to-use 10% (w/v) solution of purified NP-40 detergent for use in routine and high-demand protein research methods and molecular biology techniques. NP-40 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins. Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Properties of NP-40 Detergent:
• Molecular Weight: 617g
• Detergent Class: Nonionic
• Aggregation Number: 149
• Micelle Molecular Weight: 90,000g
• Critical Micelle Concentration (CMC): 0.29 mM (0.0179%, w/v)
• Cloud Point: 80°C
• Dialyzable: No

Specifications for NP-40 Surfact-Amps Detergent Solution:
• Visual: Clear, colorless solution, free of particulate matter.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0μeq/mL
• Carbonyls: ≤1.0μeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
HDPE bottle
Form
Liquid
Product Line
Surfact-Amps™
Reagent Type
Detergent Solution
Quantity
500 mL

Contents & Storage

Store between 20°C and 25°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text