Thermo Scientific™
VISIONlite™ Spectrophotometer Software
Catalog number: 869-191600
Have Questions?
Thermo Scientific™
VISIONlite™ Spectrophotometer Software
Catalog number: 869-191600
Have Questions?
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ ColorCalc software is a powerful stand-alone software package that adds the ability to perform numerous color calculations for a variety of applications. The software records the required transmission or reflectance spectrum and automatically performs calculations using the recorded data.
Thermo Scientific™ VISIONlite™ EnzLab software offers predefined methods and operation procedures for all standard enzymatic test kits including Citric Acid, Lactic Acid, D-Glucose, D-Fructose, Ethanol, Lactose and many more. EnzLab automates and facilitates enzymatic determinations, ensures correct calculations, and provides appropriate analytical reports.
Thermo Scientific™ VISIONlite™ Wine Analysis software allows users to perform an extensive list of predefined photometric, colorimetric and enzymatic analytical procedures common in wine and juice analysis. This PC software features a simple user interface, reliable data recording, and error-free evaluation of results within comprehensive reports.
Catalog Number
869-191600
Unit Size
Each
Description
Stand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for enzymatic test kit analysis.
Price (USD)
Full specifications
DescriptionStand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for enzymatic test kit analysis.
TypeVISIONlite EnzLab Software
For Use With (Equipment)Thermo Scientific GENESYS 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean.
Required AccessoriesInterface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Easy to use and intuitive interface for wavelength scanning, kinetics, single or multiple fixed wavelength measurements, and additional quantitative analysis controls
Price (USD)
Full specifications
DescriptionEasy to use and intuitive interface for wavelength scanning, kinetics, single or multiple fixed wavelength measurements, and additional quantitative analysis controls
TypeVISIONlite 5 Software + Cable
For Use With (Equipment)Thermo Scientific GENESYS 40-180 Spectrophotometers
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean
Required AccessoriesInterface cable included for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Scan and calculate color parameters Tristimulus (XYZ) CIE L*a*b* Multiple illuminants 2° and 10° observer angles
Price (USD)
Full specifications
DescriptionScan and calculate color parameters Tristimulus (XYZ) CIE L*a*b* Multiple illuminants 2° and 10° observer angles
TypeVISIONlite ColorCalc Software
For Use With (Equipment)Thermo Scientific GENESYS 30, 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean
Required AccessoriesInterface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers. 085-820200 - Standard USB cable for Thermo Scientific GENESYS 30 Spectrophotometer.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Stand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for wine and juice analysis.
Price (USD)
Full specifications
DescriptionStand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for wine and juice analysis.
TypeVISIONlite Wine Analysis Software
For Use With (Equipment)Thermo Scientific GENESYS 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean.
Required AccessoriesInterface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers.
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
869-191600Full specifications
EachStand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for enzymatic test kit analysis.Request A Quote
DescriptionStand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for enzymatic test kit analysis.
TypeVISIONlite EnzLab Software
For Use With (Equipment)Thermo Scientific GENESYS 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean.
Required AccessoriesInterface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers.
Unit SizeEach
840-312300Full specifications
EachEasy to use and intuitive interface for wavelength scanning, kinetics, single or multiple fixed wavelength measurements, and additional quantitative analysis controlsRequest A Quote
DescriptionEasy to use and intuitive interface for wavelength scanning, kinetics, single or multiple fixed wavelength measurements, and additional quantitative analysis controls
TypeVISIONlite 5 Software + Cable
For Use With (Equipment)Thermo Scientific GENESYS 40-180 Spectrophotometers
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean
Required AccessoriesInterface cable included for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers
Unit SizeEach
869-175600Full specifications
EachScan and calculate color parameters Tristimulus (XYZ) CIE L*a*b* Multiple illuminants 2° and 10° observer anglesRequest A Quote
DescriptionScan and calculate color parameters Tristimulus (XYZ) CIE L*a*b* Multiple illuminants 2° and 10° observer angles
TypeVISIONlite ColorCalc Software
For Use With (Equipment)Thermo Scientific GENESYS 30, 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean
Required AccessoriesInterface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers. 085-820200 - Standard USB cable for Thermo Scientific GENESYS 30 Spectrophotometer.
Unit SizeEach
869-191700Full specifications
EachStand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for wine and juice analysis.Request A Quote
DescriptionStand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for wine and juice analysis.
TypeVISIONlite Wine Analysis Software
For Use With (Equipment)Thermo Scientific GENESYS 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers
IncludesDVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean.
Required AccessoriesInterface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers.
Unit SizeEach
Showing 4 of 4
VISIONlite Software enhancements include:
Scan:
  • Peak pick to identify and label wavelength peaks or valleys
  • Import, save or export data in native or CSV format for easy archiving and reporting
  • Auto scaling of data plus simple zoom controls for detailed examination of spectra 
Rate:  For SPECTRONIC 200 and GENESYS 30, measure multi-cell kinetics in parallel or serial mode
  • Automatic enzyme activity calculation
  • Automatic data storage plus import or export your data in CSV format
  • Measure up to 100 samples in a single data set 
Fixed:
  • Measure at one or multiple wavelengths
  • Apply factors and equations to your data to generate results directly
  • Addition, subtraction, ratio and baseline correction options
  • Run standard bio-tests using the preprogrammed protein and DNA methods
Quant: More standards and non-linear curve fitting options than can be accomplished under local control on Thermo Scientific SPECTRONIC 200 and GENESYS 30 Spectrophotometers