Thermo Scientific™

Pierce™ D-Luciferin, Monosodium Salt

Catalog number: 88292
Have Questions?
Thermo Scientific™

Pierce™ D-Luciferin, Monosodium Salt

Catalog number: 88292
Have Questions?
Catalog Number
88292
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
882921 g
-
88293100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce D-Luciferin, Monopotassium Salt is formulated at greater than 99% purity to provide convenience and high performance at a low cost in reporter assays involving firefly luciferase.

Features of D-Luciferin, Monopotassium Salt:

• Molecular mass = 318.42 g/mol
• High purity, typically greater than 99.7%, as verified by HPLC and FTIR
• Great performance with in vitro applications
• Suitable for in vivo imaging

Luciferin is the substrate for bioluminescence reactions with firefly luciferase enzyme. Lyophilized potassium salt of firefly luciferin is stable and easily soluble, and is compatible with a wide variety of luciferase-based in vivo imaging and in vitro bioluminescent assays. Applications include ATP assays, gene reporter assays, high-throughput sequencing and various contamination assays throughout the biotechnology field. For in vivo imaging, the injection of luciferin in model systems enables real-time, non-invasive monitoring of disease progression and/or drug efficacy through bioluminescence.

Applications:
• Gene reporter assays
• ATP assays
• High throughput sequencing
• Contamination assays
• Bioluminescent in vivo imaging
• Drug screening
• Multiplexed assays to study off-target effects
• Signal transduction pathway analysis

Related Products
Pierce™ D-Luciferin, Monosodium Salt
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Bioluminescence
For Use With (Application)
ATP Assay, Gene Reporter Assay
Product Type
D-Luciferin, Monosodium Salt
Product Line
Pierce™
Quantity
1 g
Substrate Properties
Chemical Substrate
Substrate Type
Luciferase Substrate

Contents & Storage

Store at or below -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text