Pierce™ D-Luciferin, Monopotassium Salt
Pierce™ D-Luciferin, Monopotassium Salt
Thermo Scientific™

Pierce™ D-Luciferin, Monopotassium Salt

Thermo Scientific Pierce D-Luciferin, Monopotassium Salt is formulated at greater than 99% purity to provide convenience and high performance atRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
88293100 mg
882921 g
Catalog number 88293
Price (USD)
90.65
Online Exclusive
106.00
Save 15.35 (14%)
Each
-
Add to cart
Quantity:
100 mg
Price (USD)
90.65
Online Exclusive
106.00
Save 15.35 (14%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce D-Luciferin, Monopotassium Salt is formulated at greater than 99% purity to provide convenience and high performance at a low cost in reporter assays involving firefly luciferase.

Features of D-Luciferin, Monopotassium Salt:

• Molecular mass = 318.42 g/mol
• High purity, typically greater than 99.7%, as verified by HPLC and FTIR
• Great performance with in vitro applications
• Suitable for in vivo imaging

Luciferin is the substrate for bioluminescence reactions with firefly luciferase enzyme. Lyophilized potassium salt of firefly luciferin is stable and easily soluble, and is compatible with a wide variety of luciferase-based in vivo imaging and in vitro bioluminescent assays. Applications include ATP assays, gene reporter assays, high-throughput sequencing and various contamination assays throughout the biotechnology field. For in vivo imaging, the injection of luciferin in model systems enables real-time, non-invasive monitoring of disease progression and/or drug efficacy through bioluminescence.

Applications:
• Gene reporter assays
• ATP assays
• High throughput sequencing
• Contamination assays
• Bioluminescent in vivo imaging
• Drug screening
• Multiplexed assays to study off-target effects
• Signal transduction pathway analysis

Related Products
Pierce™ D-Luciferin, Monosodium Salt
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)ATP Assay, Gene Reporter Assay
Product LinePierce™
Product TypeD-Luciferin, Monopotassium Salt
Quantity100 mg
Substrate PropertiesChemical Substrate
Substrate TypeLuciferase Substrate
Detection MethodBioluminescence
Unit SizeEach
Contents & Storage
Store at or below -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3225875PRODUCT DATA SHEETJul 01, 202588293
3215648PRODUCT DATA SHEETJun 24, 202588292
3212123APRODUCT DATA SHEETJun 09, 202588293
3212123PRODUCT DATA SHEETMay 28, 202588293
AA390587Certificate of AnalysisJan 31, 202588292
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection