Thermo Scientific™

Pierce™ Reserpine Standard for LC-MS

Catalog number: 88326
Have Questions?
Thermo Scientific™

Pierce™ Reserpine Standard for LC-MS

Catalog number: 88326
Have Questions?
Catalog Number
88326
Unit Size
5 x 1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
883265 x 1 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Reserpine Standard for LC-MS is a precise concentration of reserpine specifically designed for performance evaluation of mass spectrometers, including the Thermo Scientific ion trap and TSQ series of instruments.

Features of Reserpine Standard for LC-MS:

Convenient—provided pre-diluted and requires minimal additional dilutions to reach target concentration for injection
Safe handling—unlike glass ampules, cap can be easily removed and replaced when withdrawing solution
High-purity—mass spectrometry grade reagent in a non-leachable screw-cap vials
Stable—store at 4°C for up to one year

Pierce Reserpine Standard for LC-MS is a pre-diluted liquid formulation ideal to quickly perform installation specification tests of Thermo Scientific and other manufacturers' mass spectrometers. The standard is provided in a concentration of 100pg/μL in 50% isopropyl alcohol and requires minimal additional dilutions. The product is provided in a pack of 5 x 1 mL glass amber vials with PTFE-lined screw caps. Pierce Reserpine Standard for LC-MS is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications.

Pierce Reserpine Standard for LC-MS can be used to assess performance of mass spectrometers, including the Thermo Scientific ion trap series and the TSQ series.
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Mass Spectrometer
Product Type
Reserpine Standard
Product Line
Pierce™
Starting Material
Peptides, Small Molecules
Final Product Type
Peptides, Small Molecules
Detection Method
Mass Spectrometry
Workflow Step
Calibration
Form
Liquid

Contents & Storage

Store in a cool, dry, well-ventilated area, protected from direct sunlight.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text