Thermo Scientific™

Pierce™ Triple Quadrupole Calibration Solution, extended mass range

Catalog number: 88340
Have Questions?
Thermo Scientific™

Pierce™ Triple Quadrupole Calibration Solution, extended mass range

Catalog number: 88340
Have Questions?
Catalog Number
88340
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
8834010 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution, Extended Mass Range, is a mixture of 14 highly pure, ionizable components (mass range: 69 m/z to 2800m/z) designed for both positive and negative ionization calibration of Thermo Scientific Triple Stage Quadrupole instruments.

Improved mass accuracy—extended mass range designed to improve mass accuracy when compared to existing calibration solutions
Positive and negative mode calibration—single formulation with extended mass ranges for both ionization calibration modes
Highly validated—high-quality, rigorously tested formulation with lot-specific certificates of analysis
Ready to use—reference standard may be directly loaded into a syringe and injected into the instrument

The Pierce Triple Quadrupole Calibration Solution is a ready-to-use formulation that has been optimized for both positive and negative ionization calibration of instruments. The extended range masses (69 m/z to 2800m/z) improve performance of the Thermo Scientific Quantiva™ and Endura™ Triple Quadrupole mass spectrometers. Additionally, the Triple Quadrupole Calibration Solution, Extended Mass Range, is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.

The Pierce Triple Quadrupole Calibration Solution, Extended Mass Range, formulation contains 14 compounds that are ionizable in both positive and negative modes for instrument calibration. The formulation has been specifically optimized for use with the TSQ Endura™ Series and TSQ Quantiva™ Series instruments. The calibration solution is also compatible with the TSQ Quantum™ Series#, TSQ Discovery™ Series#, TSQ Quantum Ultra™ Series#, TSQ Quantum Access™ Series#, and TSQ Vantage™ Series# instruments using the list of recommended masses.

Positive mode (m/Z): 69.0447, 142.1590, 322.0481, 622.0289, 922.0098
Negative mode (m/Z): 59.0128, 112.9845, 301.9970, 601.9779, 1033.9870

For the calibration procedure use:
TSQ Series Getting Started Guide (70111-97152)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Mass Spectrometer
Product Type
Calibration Solution
Product Line
Pierce™
Starting Material
Peptides, Small Molecules
Final Product Type
Peptides, Small Molecules
Detection Method
Mass Spectrometry
Workflow Step
Calibration
Form
Liquid

Contents & Storage

1 x 10 mL
Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text