Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Use With (Equipment) | Microplate Reader |
Product Type | Peptide Assay |
Product Line | Pierce™ |
Starting Material | Peptides, Protease-digested Protein |
Detection Method | Fluorescent |
Workflow Step | In-Process Testing |
Quantity | 25 x 96-well Plates |
Unit Size | 25 plates |
Catalog Number | Specifications | Unit Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
88378 | 25 plates | 160.65 Online price 172.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Thermo Scientific Pierce Quantitative Fluorescent Peptide Assay is a sensitive, mix-and-read fluorescent microplate assay for the quantitative measurement of peptides and peptide mixtures.
Because of the labeling mechanism of the fluorescent assay reagent, this assay is suitable for the quantitative measurement of synthetic peptides as well as peptide digest mixtures. A high quality peptide digest reference standard is included for use in generating linear standard curves and as a calibration control for improved accuracy and reproducibility. This sensitive assay requires only 10 μL of sample, produces a linear response with increasing peptide concentrations (5 to 1000 μg/mL), and results in a stable final fluorescence that can be detected in as little as five minutes.
Applications: