Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific Pierce Peptide Desalting Spin Columns are ready-to-use centrifuge spin columns that enable efficient desalting of peptide samples following enzymatic digestion. The polymer-based hydrophobic resin contained in each column provides excellent binding and recovery characteristics for peptide samples in preparation for mass spectrometry and other methods. Each spin column can bind from 5 μg to 5 mg of native and TMT-labeled peptides. The spin column format allows processing of multiple samples (10–300 μL each) in parallel in approximately 30 minutes.
Product Type | Spin Desalting Column |
Product Line | Pierce™ |
Shipping Condition | Room Temperature |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Format | Spin Column |
Quantity | 25 Columns |
Unit Size | 25 columns |
Product Type | Spin Desalting Column |
Product Line | Pierce™ |
Shipping Condition | Room Temperature |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Format | Spin Column |
Quantity | 50 Columns |
Unit Size | 50 columns |
Catalog Number | Specifications | Unit Size | Product Type | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
89852 | 25 columns | Spin Desalting Column | 272.65 Online price 288.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
89851 | 50 columns | Spin Desalting Column | 441.65 Online price 470.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|