Thermo Scientific™

Pierce™ Centrifuge Columns, 0.8 mL

Catalog number: 89869
Have Questions?
Thermo Scientific™

Pierce™ Centrifuge Columns, 0.8 mL

Catalog number: 89869
Have Questions?
Catalog Number
89869
Unit Size
4 x 50 columns
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
Custom
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
50 columns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
898694 x 50 columns
-
89869BCustom
--
8986850 columns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Centrifuge Columns are convenient tools for handling 40 μL to 10 mL of an affinity support for purification.

Column details:

• Made from low protein binding polypropylene for compatibility with protein purification
• Total volume: 800 μL
• Resin volume: 40-400 μL
• Filter type, pore size: Polyethylene filter, ∼30 μm pore size
• Receiver tube: Fits standard microcentrifuge tubes
• Caps: O-ring screw-top cap
• Twist-off bottom

Simply add the affinity resin to one of the polypropylene columns, remove the twist-off bottom and allow the resin to pack itself. Then add your sample and allow it to bind to the support and use a centrifuge to efficiently wash away any contaminants and elute your purified protein. Pierce Centrifuge Columns can be used in either spin or gravity-flow format. The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and when slow processing time is desired for binding steps with dilute samples, Column Extenders can be used to increase the column reservoir capacity. Use the any combination of processing techniques to get the best possible chromatography results.

Applications:

• Affinity purification, affinity chromatography
• Immunodepletion
• Ion exchange resins
• Gel filtration, desalting, buffer exchange

Related Products
Pierce™ Centrifuge Columns, 2 mL
Pierce™ Centrifuge Columns, 5 mL
Pierce™ Centrifuge Columns, 10 mL
Column Extenders for Pierce™ Centrifuge Columns
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
0.8 mL
Product Line
Pierce™
Quantity
4 x 50 Columns
Type
Spin Column
Format
Microcentrifuge, Spin Column
Column Type
Empty Column, Polyethylene Filter

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text