1x1 image pixel for data collection
CHCA MALDI Matrix, Single-Use
CHCA MALDI Matrix, Single-Use
Thermo Scientific™

CHCA MALDI Matrix, Single-Use

The Thermo Scientific CHCA MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing upRead more
Have Questions?
Catalog NumberQuantity
9003124 x 1 mg
Catalog number 90031
Price (USD)
232.90
Special offer
Online exclusive
Ends: 15-Aug-2025
274.00
Save 41.10 (15%)
Each
-
Add to cart
Quantity:
24 x 1 mg
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
232.90
Special offer
Online exclusive
Ends: 15-Aug-2025
274.00
Save 41.10 (15%)
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific CHCA MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.

Features of CHCA MALDI Matrices:

• Single-use packaging saves time and reduces waste
• Provides accurate quantity of matrix every time
• Applications: peptides, lipids, nucleotides

Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The unique packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA) MALDI matrices makes it easy to prepare high-quality MALDI reagents in minutes.

MALDI and electrospray ionization (ESI) are the most common methods to ionize peptides for MS analysis. MALDI is widely used because it is significantly faster than ESI and the ionized molecules are almost exclusively singly charged, making interpretation easier. Also, MALDI is readily automated and highly sensitive. This powerful method for MS peptide analysis is now easier to perform with our single-use, highly pure MALDI matrices that are exceptionally convenient to use.

Related Products
MALDI Matrix Sample Pack, Single-Use
SA MALDI Matrix, Single-Use
DHB MALDI Matrix, Single-Use
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce CHCA MALDI Matrix
Final Product TypeSmall Molecules
For Use With (Equipment)Mass Spectrometer
Quantity24 x 1 mg
Workflow StepCalibration
Detection MethodMass Spectrometry
FormSolid
Product TypeMALDI Matrix
Starting MaterialSmall Molecules
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°C.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3213733Certificate of AnalysisJun 02, 202590031
AB405907Certificate of AnalysisMar 20, 202590031
ZK401333ACertificate of AnalysisJan 31, 202590031
ZK401333Certificate of AnalysisJan 30, 202590031
ZJ400643Certificate of AnalysisOct 18, 202490031
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection