Thermo Scientific™

iodoTMTsixplex™ Isobaric Label Reagents and Kits

Catalog number: 90100
Have Questions?
Thermo Scientific™

iodoTMTsixplex™ Isobaric Label Reagents and Kits

Catalog number: 90100
Have Questions?
Thermo Scientific iodoTMT Label reagents enable multiplexed identification and quantitation of cysteine containing peptides in different samples using tandem mass spectrometry. IodoTMT reagents are sets of isobaric (mass and structure) isomers that are iodoacetyl-activated for covalent, irreversible labeling of sulfhydryl (–SH) groups. iodoTMTzero Label Reagent allows testing and optimization of sample preparation, labeling, fractionation, and MS fragmentation for peptide identification and reporter detection without using the more costly isotope-labeled compounds.
 
Catalog Number
90100
Unit Size
5 x 0.2 mg
Description
iodoTMTzero™ Label Reagent, 5 x 0.2 mg
Price (USD)
Availability
-
Full specifications
Quantity
DescriptioniodoTMTzero™ Label Reagent, 5 x 0.2 mg
For Use With (Equipment)Mass Spectrometer
Product LineiodoTMTzero™
Label or DyeTandem Mass Tag™ (TMT) Labels
Starting MaterialCell Lysate
Final Product TypeProtein
Detection MethodMass Spectrometry
Workflow StepProtein Labeling
Quantity5 x 0.2 mg
Unit Size5 x 0.2 mg
Catalog Number
Unit Size
1 x 0.2 mg (per tag)
Description
iodoTMTsixplex™ Isobaric Label Reagent Set, 1 x 0.2 mg
Price (USD)
Availability
-
Full specifications
Quantity
DescriptioniodoTMTsixplex™ Isobaric Label Reagent Set, 1 x 0.2 mg
For Use With (Equipment)Mass Spectrometer
Product LineiodoTMTsixplex™
Label or DyeTandem Mass Tag™ (TMT) Labels
Starting MaterialCell Lysate
Final Product TypeProtein
Detection MethodMass Spectrometry
Workflow StepProtein Labeling
Quantity1 x 0.2 mg (per tag)
Unit Size1 x 0.2 mg (per tag)
Catalog Number
Unit Size
5 x 0.2 mg (per tag)
Description
iodoTMTsixplex™ Isobaric Label Reagent Set, 5 x 0.2 mg
Price (USD)
Availability
-
Full specifications
Quantity
DescriptioniodoTMTsixplex™ Isobaric Label Reagent Set, 5 x 0.2 mg
For Use With (Equipment)Mass Spectrometer
Product LineiodoTMTsixplex™
Label or DyeTandem Mass Tag™ (TMT) Labels
Starting MaterialCell Lysate
Final Product TypeProtein
Detection MethodMass Spectrometry
Workflow StepProtein Labeling
Quantity5 x 0.2 mg (per tag)
Unit Size5 x 0.2 mg (per tag)
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
90100Full specifications
5 x 0.2 mgiodoTMTzero™ Label Reagent, 5 x 0.2 mg
-
DescriptioniodoTMTzero™ Label Reagent, 5 x 0.2 mg
For Use With (Equipment)Mass Spectrometer
Product LineiodoTMTzero™
Label or DyeTandem Mass Tag™ (TMT) Labels
Starting MaterialCell Lysate
Final Product TypeProtein
Detection MethodMass Spectrometry
Workflow StepProtein Labeling
Quantity5 x 0.2 mg
Unit Size5 x 0.2 mg
90101Full specifications
1 x 0.2 mg (per tag)iodoTMTsixplex™ Isobaric Label Reagent Set, 1 x 0.2 mg
-
DescriptioniodoTMTsixplex™ Isobaric Label Reagent Set, 1 x 0.2 mg
For Use With (Equipment)Mass Spectrometer
Product LineiodoTMTsixplex™
Label or DyeTandem Mass Tag™ (TMT) Labels
Starting MaterialCell Lysate
Final Product TypeProtein
Detection MethodMass Spectrometry
Workflow StepProtein Labeling
Quantity1 x 0.2 mg (per tag)
Unit Size1 x 0.2 mg (per tag)
90102Full specifications
5 x 0.2 mg (per tag)iodoTMTsixplex™ Isobaric Label Reagent Set, 5 x 0.2 mg
-
DescriptioniodoTMTsixplex™ Isobaric Label Reagent Set, 5 x 0.2 mg
For Use With (Equipment)Mass Spectrometer
Product LineiodoTMTsixplex™
Label or DyeTandem Mass Tag™ (TMT) Labels
Starting MaterialCell Lysate
Final Product TypeProtein
Detection MethodMass Spectrometry
Workflow StepProtein Labeling
Quantity5 x 0.2 mg (per tag)
Unit Size5 x 0.2 mg (per tag)
Showing 3 of 3

Iodoacetyl tandem mass tag reagents enable selective labeling of cysteine-containing peptides for multiplexed sample quantitation using LC-MS/MS analysis. Similar to iodoacetamide, iodoTMT reagents react specifically with reduced cysteines (Cys) in peptides and proteins and enable quantitation of the relative abundance of cysteine modifications, such as S-nitrosylation and the oxidation of disulfide bonds. IodoTMT reagents irreversibly label free sulfhydryl groups on cysteine residues, in contrast to previous reversible cysteine-reactive TMT (cysTMT) reagents.

To selectively analyze the cysteine-labeled peptides, the sample may be enriched using Immobilized Anti-TMT Antibody Resin (Cat. No. 90076) and TMT Elution Buffer (Cat. No. 90104). This approach of selective thiol-labeling and affinity enrichment is similar to isotope-coded affinity tags (ICAT method) but allows for affinity enrichment with a non-biological tag and higher multiplex quantitation.

Features of iodoTMT reagents
Specific—only reacts with sulfhydryl groups (reduced, unmodified cysteine residues)
Irreversible—labeled proteins and peptides are not susceptible to reducing agents
Flexible—options for duplex isotopic (MS) or sixplex isobaric (MS/MS) quantitation
Versatile—can be used to study cysteine modifications (e.g., di-sulfide bridges, S-nitrosylation)

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

General References:

Thompson, A., et al. (2003). Tandem mass tags: a novel quantification strategy for comparative analysis of complex protein mixtures by MS/MS. Anal. Chem. 75:1895-204.

Forrester MT, et al. (2009). Detection of protein S-nitroylsation with the biotin technique. Free Radic Biol Med. 46(2):119-126.

Gygi, S.P., et al. (1999). Quantitative analysis of complex protein mixtures using isotopecoded affinity tags. Nat. Biotech. 17:994-9.

Murray CI, et al. (2012) Identification and quantification of S-nitrosylation by cysteine reactive tandem mass tag switch assay. Mol Cell Proteomics. 11(2): M111.013441.

Yao, C., et al. (2013). Proteomic study of reversible cysteine oxidation in catalase overexpressing mice on a western diet. FASEB J, Apr 2013; 27:810.3.

Contents & Storage

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text